Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other NFIB SIRNA Assays ›
          Gene Symbol:
          NFIB
          Gene Name:
          nuclear factor I B
          Gene Aliases:
          CTF, HMGIC/NFIB, NF-I/B, NF1-B, NFI-B, NFI-RED, NFIB2, NFIB3
          Chromosome Location:
          Chr.9: 14081843 - 14398983 on Build GRCh38
          UniGene ID:
          Hs.644095
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s9494

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: NFIB
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001190737.1 NP_001177666.1 7, 8 1600
          NM_001190738.1 NP_001177667.1 7, 8 1495
          NM_001282787.1 NP_001269716.1 6, 7 647
          NM_005596.3 NP_005587.2 7, 8 1600
          XM_005251467.2 XP_005251524.1 7, 8 1693
          XM_005251468.4 XP_005251525.1 7, 8 1166
          XM_005251469.2 XP_005251526.1 7, 8 1693
          XM_005251470.2 XP_005251527.1 7, 8 1693
          XM_005251471.3 XP_005251528.1 6, 7 2528
          XM_006716774.2 XP_006716837.1 7, 8 1693
          XM_006716775.2 XP_006716838.1 7, 8 1693
          XM_017014740.1 XP_016870229.1 7, 8 1511
          XM_017014742.1 XP_016870231.1 6, 7 3054
          GenBank mRNA AK131233.1 - 7, 8 1495
          AK290603.1 - 7, 8 1213
          AK299605.1 - 7, 8 1463
          BC001283.1 - 7, 8 1223 3462726
          BT007266.1 - 7, 8 1065
          BX648416.1 - 6, 7 644
          BX648845.1 - 7, 8 1600
          KU178232.1 - 7, 8 1065
          KU178233.1 - 8, 9 1143
          U85193.1 - 7, 8 1274

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_031566, XM_008763762, XM_006238335, XM_006238336, XM_006238339, and others.
          This siRNA also targets the following transcripts in Mouse: NM_001113209, NM_001286131, NM_001113210, NM_008687, NM_001286127, and others.

          Panther Classification:

          Molecular Function -

          DNA-binding transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          chondrocyte differentiation
          DNA replication
          transcription from RNA polymerase II promoter
          glial cell differentiation
          principal sensory nucleus of trigeminal nerve development
          anterior commissure morphogenesis
          hindbrain development
          positive regulation of transcription from RNA polymerase II promoter
          Clara cell differentiation
          Type I pneumocyte differentiation
          Type II pneumocyte differentiation
          salivary gland cavitation
          cell differentiation involved in salivary gland development
          lung ciliated cell differentiation
          commissural neuron axon guidance
          negative regulation of pri-miRNA transcription from RNA polymerase II promoter
          negative regulation of mesenchymal cell proliferation involved in lung development
          negative regulation of epithelial cell proliferation involved in lung morphogenesis
          RNA polymerase II core promoter proximal region sequence-specific DNA binding
          RNA polymerase II transcription factor activity, sequence-specific DNA binding
          transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
          DNA binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0