Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Epm2aip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175266.4 | NP_780475.1 | 1 | 1398 | ||
GenBank mRNA | AK030084.1 | - | 1 | 1315 | ||
AK032284.1 | - | 1 | 1369 | |||
AK043518.1 | - | 1 | 1357 | |||
AK122366.1 | - | 1 | 491 | |||
BC018474.1 | - | 1 | 1402 | 3156440 |