Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACTG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199954.1 | NP_001186883.1 | 6 | 1495 | ||
NM_001614.3 | NP_001605.1 | 6 | 1376 | |||
NR_037688.1 | 6 | 1376 | ||||
GenBank mRNA | AK291937.1 | - | 6 | 1309 | ||
AK301175.1 | - | 7 | 1183 | |||
AK304637.1 | - | 7 | 1249 | |||
BC000292.2 | - | 6 | 1302 | 2819345 | ||
BC001920.1 | - | 6 | 1265 | 2820489 | ||
BC004223.2 | - | 5 | 1214 | |||
BC007442.2 | - | 6 | 1296 | 2962953 | ||
BC009544.1 | - | 3 | 590 | |||
BC009848.2 | - | 6 | 1293 | 3939552 | ||
BC010417.2 | - | 3 | 609 | |||
BC010999.2 | - | 6 | 1295 | 3940861 | ||
BC012050.1 | - | 6 | 1296 | 4554944 | ||
BC015005.1 | - | 6 | 1291 | 4855463 | ||
BC015695.2 | - | 6 | 1298 | 4842665 | ||
BC015779.1 | - | 6 | 1272 | 4856294 | ||
BC017450.1 | - | 5 | 1203 | |||
BC018774.1 | - | 6 | 1296 | 4869628 | ||
BC018861.2 | - | 3 | 736 | |||
BC023548.2 | - | 4 | 903 | |||
BC053572.1 | - | 6 | 1289 | 3865767 | ||
BC063495.1 | - | 5 | 1228 | |||
EU668331.1 | - | 3 | 808 | |||
M16247.1 | - | 3 | 808 | |||
X04098.1 | - | 6 | 1310 |
Gene Symbol: ACTG1P20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033926.1 | 1 | 2034 | |||
GenBank mRNA | AK056682.1 | - | 1 | 2034 | ||
BX648289.1 | - | 1 | 3512 |