Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ncapd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178113.3 | NP_835214.2 | 8 | 1311 | ||
GenBank mRNA | AK014555.1 | - | 7 | 1100 | ||
AK129046.1 | - | 7 | 837 | |||
AK139081.1 | - | 7 | 869 | |||
BC048190.1 | - | 8 | 1311 | 6831876 | ||
DQ340802.1 | - | 7 | 966 |