Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other APOBEC3 SIRNA Assays ›
          Gene Symbol:
          Apobec3
          Gene Name:
          apolipoprotein B mRNA editing enzyme, catalytic polypeptide 3
          Gene Aliases:
          Arp3, BC003314, Cem15, Rfv3
          Chromosome Location:
          Chr.15: 79892408 - 79908905 on Build GRCm38
          UniGene ID:
          Mm.284059
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID s95946

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Apobec3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001160415.1 NP_001153887.1 8 1288
          NM_030255.3 NP_084531.2 7 1189
          GenBank mRNA AB461446.1 - 8 1179
          AB461447.1 - 7 1080
          AB461448.1 - 7 1080
          AB461449.1 - 7 1080
          AK044394.1 - 8 1248
          AK049256.1 - 7 1149
          AK049998.1 - 1143
          AK153719.1 - 7 1139
          AK154035.1 - 1293
          BC003314.1 - 8 1270 3155422
          BC007182.1 - 3 1425
          EU707568.1 - 8 1179
          EU707569.1 - 7 1011
          EU707570.1 - 7 1080
          EU707571.1 - 7 1011
          GQ871501.1 - 8 1176
          GQ871502.1 - 6 1020
          GQ871503.1 - 7 1077

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          RNA processing factor

          Gene Ontology Categories:

          Function(s) Process(es)

          hematopoietic progenitor cell differentiation
          immune system process
          metabolic process
          negative regulation of transposition
          negative regulation of viral genome replication
          innate immune response
          defense response to virus
          catalytic activity
          GTP cyclohydrolase activity
          protein binding
          tRNA-specific adenosine deaminase activity
          zinc ion binding
          hydrolase activity
          hydrolase activity, acting on carbon-nitrogen (but not peptide) bonds, in cyclic amidines
          N-cyclopropylmelamine deaminase activity
          N-cyclopropylammeline deaminase activity
          N-cyclopropylammelide alkylamino hydrolase activity
          2,5-diamino-6-ribitylamino-4(3H)-pyrimidinone 5'-phosphate deaminase activity
          tRNA-specific adenosine-37 deaminase activity
          archaeal-specific GTP cyclohydrolase activity
          poly(A) RNA binding
          metal ion binding
          deoxycytidine deaminase activity
          tRNA-specific adenosine-34 deaminase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0