Search
Search

Slc16a3| Gene Symbol: Slc16a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001038653.1 | NP_001033742.1 | 4 | 531 | ||
| NM_001038654.1 | NP_001033743.1 | 3 | 436 | |||
| NM_030696.3 | NP_109621.1 | 4 | 543 | |||
| GenBank mRNA | AF178954.1 | - | 2 | 331 | ||
| AK141381.1 | - | 3 | 433 | |||
| AK149707.1 | - | 4 | 534 | |||
| AK149983.1 | - | 4 | 534 | |||
| AK165596.1 | - | 3 | 379 | |||
| AK166048.1 | - | 519 | ||||
| AK169917.1 | - | 4 | 493 | |||
| AK171260.1 | - | 4 | 519 | |||
| BC046525.1 | - | 4 | 543 | 5319754 | ||