siRNA Details
Gene Symbol: NOTCH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017617.4 | NP_060087.3 | 22, 23 | 3651 | ||
XM_011518717.2 | XP_011517019.2 | 19, 20 | 2943 | |||
GenBank mRNA | AB209873.1 | - | 14, 15 | 2188 | ||
AF308602.1 | - | 22, 23 | 3654 | |||
M73980.1 | - | 22, 23 | 3654 |

More Information
Panther Classification:
Molecular Function -




