Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbm5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_148930.3 | NP_683732.1 | 20 | 1981 | ||
GenBank mRNA | AJ309168.1 | - | 19 | 1816 | ||
AK134183.1 | - | 20 | 1976 | |||
AK146880.1 | - | 20 | 1976 | |||
BC003988.1 | - | 10 | 930 | |||
BC005719.1 | - | 15 | 1586 | |||
BC023854.1 | - | 20 | 1966 | 5346780 | ||
BC031899.1 | - | 20 | 1966 | 5362121 | ||
BC042419.1 | - | 18 | 5217 | |||
BC043058.1 | - | 20 | 1918 | 6419695 | ||
BC054729.1 | - | 20 | 1969 | 6419758 | ||
BC054820.1 | - | 18 | 4695 | |||
BC060985.1 | - | 19 | 1885 |