Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Setdb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163641.1 | NP_001157113.1 | 13 | 2443 | ||
NM_001163642.1 | NP_001157114.1 | 13 | 2402 | |||
NM_018877.3 | NP_061365.3 | 13 | 2405 | |||
GenBank mRNA | AF091628.1 | - | 13 | 2356 | ||
AK088590.1 | - | 6 | 1394 | |||
AK132202.1 | - | 13 | 2378 | |||
BC060173.1 | - | 13 | 2397 |