Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NOVA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002516.3 | NP_002507.1 | 4 | 1462 | ||
XM_006723230.3 | XP_006723293.1 | 5 | 1514 | |||
XM_017026838.1 | XP_016882327.1 | 4 | 1738 | |||
XM_017026839.1 | XP_016882328.1 | 4 | 1564 | |||
XM_017026840.1 | XP_016882329.1 | 3 | 1321 | |||
GenBank mRNA | AF083898.1 | - | 4 | 1462 | ||
U70477.1 | - | 4 | 1433 |