Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other NPM1 SIRNA Assays ›
          Gene Symbol:
          NPM1
          Gene Name:
          nucleophosmin
          Gene Aliases:
          B23, NPM
          Chromosome Location:
          Chr.5: 171387648 - 171410900 on Build GRCh38
          UniGene ID:
          Hs.557550
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s9678

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: NPM1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001037738.2 NP_001032827.1 1, 2 301
          NM_002520.6 NP_002511.1 1, 2 301
          NM_199185.3 NP_954654.1 1, 2 301
          XM_005265920.3 XP_005265977.1 1, 2 357
          GenBank mRNA AB042278.1 - 1, 2 56
          AB451236.1 - 1, 2 56
          AB451361.1 - 1, 2 56
          AK290652.1 - 1, 2 164
          AY347529.1 - 2, 3 191
          AY740634.1 - 1, 2 128
          AY740635.1 - 1, 2 128
          AY740636.1 - 1, 2 128
          AY740637.1 - 1, 2 128
          AY740638.1 - 1, 2 128
          AY740639.1 - 1, 2 128
          AY740640.1 - 1, 2 128
          BC002398.2 - 1, 2 143 2821577
          BC008495.1 - 1, 2 153 4276604
          BC009623.2 - 1, 2 182 3877633
          BC012566.1 - 1, 2 156 4097025
          BC014349.1 - 1, 2 152 3511037
          BC016716.1 - 1, 2 155 4068167
          BC016768.1 - 1, 2 155 4081364
          BC016824.1 - 1, 2 155 4106286
          BC020467.1 - 1, 2 142
          BC021668.1 - 1, 2 156 3930893
          BC021983.1 - 1, 2 154 4106023
          BC050628.1 - 1, 2 142 5575414
          BC107754.1 - 1, 2 157 6736004
          BP291033.1 - 1, 2 211
          BT007011.1 - 1, 2 56
          DQ303464.1 - 1, 2 152
          EF429251.1 - 1, 2 56
          KU178236.1 - 1, 2 56
          KU178237.1 - 1, 2 56
          M23613.1 - 1, 2 149
          M26697.1 - 1, 2 146
          M28699.1 - 1, 2 143
          X16934.1 - 1, 2 16

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          chaperone

          Gene Ontology Categories:

          Function(s) Process(es)

          DNA repair
          nucleosome assembly
          intracellular protein transport
          nucleocytoplasmic transport
          response to stress
          DNA damage response, signal transduction by p53 class mediator resulting in cell cycle arrest
          centrosome cycle
          signal transduction
          cell aging
          protein localization
          positive regulation of cell proliferation
          negative regulation of cell proliferation
          negative regulation of centrosome duplication
          viral process
          regulation of endodeoxyribonuclease activity
          CENP-A containing nucleosome assembly
          ribosome assembly
          negative regulation of apoptotic process
          negative regulation of protein kinase activity by regulation of protein phosphorylation
          positive regulation of translation
          positive regulation of transcription, DNA-templated
          regulation of centriole replication
          positive regulation of NF-kappaB transcription factor activity
          protein oligomerization
          regulation of endoribonuclease activity
          regulation of eIF2 alpha phosphorylation by dsRNA
          positive regulation of cell cycle G2/M phase transition
          transcription coactivator activity
          RNA binding
          protein kinase inhibitor activity
          protein binding
          protein kinase binding
          Tat protein binding
          histone binding
          protein homodimerization activity
          ribosomal large subunit binding
          ribosomal small subunit binding
          poly(A) RNA binding
          protein heterodimerization activity
          NF-kappaB binding
          unfolded protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-zjghb:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0