Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cnnm3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039551.1 | NP_001034640.1 | 3 | 1507 | ||
NM_053186.2 | NP_444416.2 | 3 | 1507 | |||
GenBank mRNA | AF216964.1 | - | 3 | 576 | ||
AK143035.1 | - | 2 | 209 | |||
BC052714.1 | - | 3 | 574 | |||
BC080275.1 | - | 3 | 934 | |||
BC108417.1 | - | 3 | 761 | |||
BC132285.1 | - | 3 | 559 | |||
BC144720.1 | - | 3 | 558 | |||
GQ225746.2 | - | 3 | 493 |