Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hrh3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133849.3 | NP_598610.1 | 3 | 923 | ||
GenBank mRNA | AK140374.1 | - | 3 | 928 | ||
AK156130.1 | - | 1 | 250 | |||
AY044153.2 | - | 3 | 750 | |||
AY142145.1 | - | 3 | 894 | |||
BC028947.1 | - | 1 | 73 | |||
BC132201.1 | - | 3 | 690 | 40130478 | ||
BC145125.1 | - | 3 | 686 |
Set Membership: |
G-protein coupled receptor |