Search
Search

Clca4b| Gene Symbol: Clca4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033199.3 | NP_001028371.1 | 11 | 1826 | ||
| GenBank mRNA | AK136515.1 | - | 11 | 1831 | ||
| BC139088.1 | - | 11 | 1820 | 8862110 | ||
| BC139089.1 | - | 11 | 1820 | 8862111 | ||
| JN652132.1 | - | 11 | 1797 | |||