Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other NTRK3 SIRNA Assays ›
Gene Symbol:
NTRK3
Gene Name:
neurotrophic receptor tyrosine kinase 3
Gene Aliases:
GP145-TrkC, TRKC, gp145(trkC)
Chromosome Location:
Chr.15: 87859749 - 88256796 on Build GRCh38
UniGene ID:
Hs.185701
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s9752
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: NTRK3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001007156.2
NP_001007157.1
11
1525
NM_001012338.2
NP_001012338.1
11
1525
NM_001320134.1
NP_001307063.1
9
1235
NM_001320135.1
NP_001307064.1
9
1126
NM_002530.3
NP_002521.2
11
1525
XM_006720543.3
XP_006720606.1
9
1834
XM_006720545.3
XP_006720608.1
9
1834
XM_006720548.3
XP_006720611.1
9
1834
XM_006720549.3
XP_006720612.1
9
1834
XM_011521637.2
XP_011519939.1
9
1834
XM_011521638.2
XP_011519940.1
9
1834
XM_017022240.1
XP_016877729.1
10
1705
XM_017022241.1
XP_016877730.1
9
1190
XM_017022242.1
XP_016877731.1
9
1834
XM_017022243.1
XP_016877732.1
9
1186
XM_017022244.1
XP_016877733.1
9
1088
XM_017022245.1
XP_016877734.1
10
1143
XM_017022246.1
XP_016877735.1
9
1244
XM_017022249.1
XP_016877738.1
9
1834
XM_017022250.1
XP_016877739.1
9
1834
XM_017022251.1
XP_016877740.1
2
205
XM_017022253.1
XP_016877742.1
9
1834
XM_017022254.1
XP_016877743.1
9
1834
GenBank mRNA
AK302504.1
-
9
1085
BC013693.1
-
9
1224
4181574
BT007291.1
-
9
1219
KJ534904.1
-
9
1219
KJ534905.1
-
9
1219
S76475.1
-
9
1234
S76476.1
-
9
1234
U05012.1
-
10
1374
Back To Top
More Information
Panther Classification:
Molecular Function -
transmembrane signal receptor
Gene Ontology Categories:
Function(s)
Process(es)
activation of MAPK activity
neuron migration
negative regulation of protein phosphorylation
positive regulation of protein phosphorylation
transmembrane receptor protein tyrosine kinase signaling pathway
heart development
circadian rhythm
positive regulation of cell proliferation
positive regulation of gene expression
peptidyl-tyrosine phosphorylation
modulation by virus of host transcription
positive regulation of cell migration
activation of protein kinase B activity
positive regulation of peptidyl-serine phosphorylation
neurotrophin signaling pathway
mechanoreceptor differentiation
positive regulation of apoptotic process
response to ethanol
protein autophosphorylation
ephrin receptor signaling pathway
neuron fate specification
response to axon injury
positive regulation of axon extension involved in regeneration
negative regulation of astrocyte differentiation
positive regulation of positive chemotaxis
response to corticosterone
positive regulation of synapse assembly
negative regulation of cell death
lens fiber cell differentiation
cellular response to retinoic acid
cochlea development
activation of GTPase activity
positive regulation of actin cytoskeleton reorganization
p53 binding
GPI-linked ephrin receptor activity
neurotrophin receptor activity
protein binding
ATP binding
neurotrophin binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-nbkcc:80/100.66.79.31:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0