Search
Search

Zbtb48| Gene Symbol: Zbtb48 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133879.2 | NP_598640.2 | 3 | 839 | ||
| GenBank mRNA | AK027912.1 | - | 2 | 1208 | ||
| AM161454.1 | - | 3 | 787 | |||
| BC005515.1 | - | 3 | 850 | 3493611 | ||
| CX202255.1 | - | 1 | 16 | |||