Search
Search

Bsdc1| Gene Symbol: Bsdc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133889.2 | NP_598650.2 | 7 | 585 | ||
| GenBank mRNA | AK037201.1 | - | 7 | 581 | ||
| AK144053.1 | - | 7 | 580 | |||
| BC021480.1 | - | 6 | 565 | |||
| BC049111.1 | - | 7 | 585 | 6415574 | ||