Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other HSD3B7 SIRNA Assays ›
          Gene Symbol:
          Hsd3b7
          Gene Name:
          hydroxy-delta-5-steroid dehydrogenase, 3 beta- and steroid delta-isomerase 7
          Gene Aliases:
          AI195443, BB098564
          Chromosome Location:
          Chr.7: 127800609 - 127803802 on Build GRCm38
          UniGene ID:
          Mm.157103
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID s97772

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Hsd3b7
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001040684.1 NP_001035774.1 7 1057
          NM_133943.2 NP_598704.2 7 1109
          GenBank mRNA AF277718.1 - 7 1045
          AK077476.1 - 6 1160
          AK150786.1 - 2 481
          AK210009.1 - 1 70
          BC006822.1 - 5 1507
          BC012954.1 - 6 909
          BC018525.1 - 5 1507
          BC132605.1 - 7 1043 40130882
          BC138589.1 - 7 1043 8861609

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          dehydratase

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of cell growth
          steroid biosynthetic process
          oxidation-reduction process
          C-3 sterol dehydrogenase (C-4 sterol decarboxylase) activity
          3-beta-hydroxy-delta5-steroid dehydrogenase activity
          aldo-keto reductase (NADP) activity
          isocitrate dehydrogenase activity
          mevaldate reductase activity
          gluconate dehydrogenase activity
          steroid dehydrogenase activity
          oxidoreductase activity
          oxidoreductase activity, acting on the CH-OH group of donors, NAD or NADP as acceptor
          epoxide dehydrogenase activity
          5-exo-hydroxycamphor dehydrogenase activity
          2-hydroxytetrahydrofuran dehydrogenase activity
          acetoin dehydrogenase activity
          phenylcoumaran benzylic ether reductase activity
          D-xylose:NADP reductase activity
          L-arabinose:NADP reductase activity
          D-arabinitol dehydrogenase, D-ribulose forming (NADP+) activity
          steroid dehydrogenase activity, acting on the CH-OH group of donors, NAD or NADP as acceptor
          steroid dehydrogenase activity, acting on the CH-CH group of donors
          (R)-(-)-1,2,3,4-tetrahydronaphthol dehydrogenase activity
          3-hydroxymenthone dehydrogenase activity
          very long-chain-3-hydroxyacyl-CoA dehydrogenase activity
          dihydrotestosterone 17-beta-dehydrogenase activity
          (R)-2-hydroxyisocaproate dehydrogenase activity
          L-arabinose 1-dehydrogenase (NADP+) activity
          L-xylulose reductase (NAD+) activity
          cholest-5-ene-3-beta,7-alpha-diol 3-beta-dehydrogenase activity
          3-ketoglucose-reductase activity
          (R)-2-hydroxyglutarate dehydrogenase activity
          D-arabinitol dehydrogenase, D-xylulose forming (NADP+) activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-zjghb:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0