Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other NR4A2 SIRNA Assays ›
          Gene Symbol:
          NR4A2
          Gene Name:
          nuclear receptor subfamily 4 group A member 2
          Gene Aliases:
          HZF-3, NOT, NURR1, RNR1, TINUR
          Chromosome Location:
          Chr.2: 156324432 - 156334683 on Build GRCh38
          UniGene ID:
          Hs.563344
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s9785

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: NR4A2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_006186.3 NP_006177.1 8 2041
          XM_005246621.3 XP_005246678.1 7 3825
          XM_005246622.3 XP_005246679.1 8 1809
          XM_006712553.3 XP_006712616.1 7 3750
          XM_011511246.1 XP_011509548.1 8 3721
          XM_017004219.1 XP_016859708.1 7 3384
          XM_017004220.1 XP_016859709.1 8 3874
          GenBank mRNA AK223625.1 - 8 1982
          AK291456.1 - 8 1802
          AK299439.1 - 7 1855
          BC009288.2 - 8 1971 4298967
          BC066890.1 - 8 1772 4828460
          HQ692824.1 - 6 1619
          HQ709176.1 - 6 1544
          KU178238.1 - 6 1619
          KU178239.1 - 6 1607
          S77154.1 - 8 1628
          X75918.1 - 8 1936

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          C4 zinc finger nuclear receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          response to hypoxia
          neuron migration
          response to amphetamine
          transcription, DNA-templated
          transcription initiation from RNA polymerase II promoter
          signal transduction
          adult locomotory behavior
          post-embryonic development
          central nervous system projection neuron axonogenesis
          habenula development
          intracellular receptor signaling pathway
          cellular response to extracellular stimulus
          cellular response to oxidative stress
          regulation of dopamine metabolic process
          dopamine biosynthetic process
          neuron maturation
          positive regulation of catalytic activity
          steroid hormone mediated signaling pathway
          negative regulation of neuron apoptotic process
          regulation of respiratory gaseous exchange
          fat cell differentiation
          positive regulation of transcription from RNA polymerase II promoter
          general adaptation syndrome
          canonical Wnt signaling pathway
          cellular response to corticotropin-releasing hormone stimulus
          dopaminergic neuron differentiation
          midbrain dopaminergic neuron differentiation
          negative regulation of apoptotic signaling pathway
          RNA polymerase II core promoter sequence-specific DNA binding
          transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
          DNA binding
          steroid hormone receptor activity
          RNA polymerase II transcription factor activity, ligand-activated sequence-specific DNA binding
          protein binding
          beta-catenin binding
          zinc ion binding
          glucocorticoid receptor binding
          protein homodimerization activity
          retinoid X receptor binding
          protein heterodimerization activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0