Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other BBS4 SIRNA Assays ›
          Gene Symbol:
          Bbs4
          Gene Name:
          Bardet-Biedl syndrome 4 (human)
          Gene Aliases:
          AW537059, AW742241, D9Ertd464e
          Chromosome Location:
          Chr.9: 59321966 - 59353508 on Build GRCm38
          UniGene ID:
          Mm.200714
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID s97998

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price 5,354.00
          Your Price
          Online offer:
          5354.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Bbs4
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_175325.3 NP_780534.1 6 462
          GenBank mRNA AK039560.1 - 6 411
          AK089970.1 - 6 417
          AK134734.1 - 6 415
          BC055797.1 - 6 382
          BC089507.1 - 6 396
          BC092531.1 - 6 406
          BC145771.1 - 6 424 40131074
          BC145773.1 - 6 424 40131076

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          microtubule cytoskeleton organization
          mitotic cytokinesis
          neural tube closure
          retina homeostasis
          negative regulation of systemic arterial blood pressure
          transport
          spermatid development
          sensory perception of smell
          protein localization
          metabolic process
          negative regulation of gene expression
          protein transport
          dendrite development
          regulation of lipid metabolic process
          ventricular system development
          striatum development
          hippocampus development
          cerebral cortex development
          cell projection organization
          adult behavior
          negative regulation of actin filament polymerization
          regulation of cytokinesis
          leptin-mediated signaling pathway
          protein localization to organelle
          negative regulation of GTPase activity
          microtubule anchoring at centrosome
          nonmotile primary cilium assembly
          social behavior
          photoreceptor cell outer segment organization
          negative regulation of appetite by leptin-mediated signaling pathway
          positive regulation of multicellular organism growth
          cilium assembly
          response to leptin
          photoreceptor cell maintenance
          positive regulation of cilium assembly
          retinal rod cell development
          brain morphogenesis
          centrosome organization
          maintenance of protein location in nucleus
          regulation of stress fiber assembly
          cilium morphogenesis
          regulation of cilium beat frequency involved in ciliary motility
          face development
          fat pad development
          protein localization to cilium
          protein localization to centrosome
          regulation of nonmotile primary cilium assembly
          protein localization to photoreceptor outer segment
          RNA polymerase II repressing transcription factor binding
          microtubule motor activity
          protein binding
          dynactin binding
          alpha-tubulin binding
          beta-tubulin binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0