Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mgat4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145926.3 | NP_666038.3 | 4 | 782 | ||
GenBank mRNA | AB053218.1 | - | 4 | 484 | ||
AK155336.1 | - | 4 | 783 | |||
AK204339.1 | - | 1 | 55 | |||
AK209484.1 | - | 2 | 151 | |||
CX205743.1 | - | 1 | 42 |