Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ctsq | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029636.3 | NP_083912.2 | 6, 7 | 866 | ||
GenBank mRNA | AF456461.1 | - | 6, 7 | 897 | ||
AK005416.1 | - | 2, 3 | 687 | |||
AY014776.1 | - | 6, 7 | 866 | |||
BC099415.1 | - | 6, 7 | 873 | 30786803 |