siRNA Details
Gene Symbol: Sncb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033610.2 | NP_291088.1 | 3 | 294 | ||
GenBank mRNA | BC019409.1 | - | 3 | 294 | 4482283 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Panther Classification:
