Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OMD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005014.2 | NP_005005.1 | 3 | 1410 | ||
GenBank mRNA | AB000114.1 | - | 3 | 1141 | ||
AK225922.1 | - | 3 | 1255 | |||
AK313055.1 | - | 3 | 1249 | |||
AY358872.1 | - | 3 | 1308 | |||
BC046356.1 | - | 3 | 1312 | 5239983 |