Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kctd9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001111028.1 | NP_001104498.1 | 11 | 1250 | ||
NM_001285933.1 | NP_001272862.1 | 11 | 1377 | |||
NM_134073.2 | NP_598834.2 | 10 | 1100 | |||
GenBank mRNA | AK087312.1 | - | 11 | 1124 | ||
AK158922.1 | - | 10 | 1102 | |||
AK166501.1 | - | 10 | 936 | |||
BC011302.1 | - | 11 | 1191 | |||
CK618608.1 | - | 4 | 395 |