Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other OPRM1 SIRNA Assays ›
Gene Symbol:
OPRM1
Gene Name:
opioid receptor mu 1
Gene Aliases:
LMOR, M-OR-1, MOP, MOR, MOR1, OPRM
Chromosome Location:
Chr.6: 154010496 - 154246867 on Build GRCh38
UniGene ID:
Hs.2353
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s9871
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: OPRM1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_000914.4
NP_000905.3
3
1535
NM_001008503.2
NP_001008503.2
3
1226
NM_001008504.3
NP_001008504.2
3
1535
NM_001008505.2
NP_001008505.2
3
1226
NM_001145279.3
NP_001138751.1
5
1894
NM_001145280.3
NP_001138752.1
3
1325
NM_001145281.2
NP_001138753.1
3
866
NM_001145282.2
NP_001138754.1
3
1226
NM_001145283.2
NP_001138755.1
3
1226
NM_001145284.3
NP_001138756.1
3
1226
NM_001145285.2
NP_001138757.1
3
1226
NM_001145286.2
NP_001138758.1
3
1226
NM_001145287.2
NP_001138759.1
3
2100
NM_001285523.1
NP_001272452.1
3
1535
NM_001285524.1
NP_001272453.1
4
1430
NM_001285526.1
NP_001272455.1
3
2081
NM_001285527.1
NP_001272456.1
2
866
NM_001285528.1
NP_001272457.1
2
866
NR_104348.1
3
1226
NR_104349.1
3
1226
NR_104350.1
2
873
NR_104351.1
3
1226
XM_011535851.2
XP_011534153.1
4
2016
XM_011535853.2
XP_011534155.1
4
1057
XM_011535856.2
XP_011534158.1
3
889
XM_011535862.2
XP_011534164.1
3
985
XM_017010903.1
XP_016866392.1
3
3010
XM_017010904.1
XP_016866393.1
3
2119
XM_017010905.1
XP_016866394.1
3
2138
XM_017010906.1
XP_016866395.1
3
3010
GenBank mRNA
AK313901.1
-
3
1275
AY036622.1
-
3
1141
AY036623.1
-
3
1141
AY195733.2
-
2
812
AY225404.1
-
3
1183
AY309001.1
-
3
1142
AY309005.1
-
3
1142
AY309006.1
-
3
1142
AY309007.1
-
3
1142
AY309008.1
-
3
1142
AY309009.1
-
3
1142
AY364230.1
-
3
1162
AY364890.1
-
2
866
AY521028.1
-
3
1092
BC074927.2
-
3
1137
30915262
EU340241.1
-
5
1889
EU340242.1
-
3
1320
EU340243.1
-
3
861
EU360599.2
-
3
2081
EU362990.2
-
3
2100
FJ041291.1
-
3
1535
FJ041292.1
-
4
1430
FJ041293.1
-
3
1140
FJ041294.1
-
3
1140
GQ258059.1
-
3
1024
HQ699462.1
-
2
788
JX914655.1
-
3
1329
L25119.1
-
3
1304
L29301.1
-
3
1330
U12569.1
-
3
1288
Back To Top
More Information
Set Membership:
G-protein coupled receptor
Panther Classification:
Molecular Function -
G-protein coupled receptor
transmembrane signal receptor
Gene Ontology Categories:
Function(s)
Process(es)
acute inflammatory response to antigenic stimulus
G-protein coupled receptor signaling pathway, coupled to cyclic nucleotide second messenger
adenylate cyclase-activating dopamine receptor signaling pathway
negative regulation of adenylate cyclase activity
phospholipase C-activating G-protein coupled receptor signaling pathway
neuropeptide signaling pathway
chemical synaptic transmission
sensory perception
locomotory behavior
negative regulation of cell proliferation
response to radiation
sensory perception of pain
adenylate cyclase-inhibiting opioid receptor signaling pathway
response to food
positive regulation of appetite
response to lipopolysaccharide
cellular response to stress
wound healing
response to cocaine
eating behavior
estrous cycle
behavioral response to ethanol
positive regulation of neurogenesis
regulation of sensory perception of pain
excitatory postsynaptic potential
negative regulation of Wnt protein secretion
positive regulation of ERK1 and ERK2 cascade
calcium ion transmembrane transport
response to growth factor
cellular response to morphine
regulation of N-methyl-D-aspartate selective glutamate receptor activity
G-protein alpha-subunit binding
G-protein coupled receptor activity
beta-endorphin receptor activity
voltage-gated calcium channel activity
protein binding
protein C-terminus binding
protein domain specific binding
filamin binding
G-protein beta-subunit binding
morphine receptor activity
neuropeptide binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0