Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bfsp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002896.2 | NP_001002896.1 | 3 | 772 | ||
GenBank mRNA | AK134810.1 | - | 3 | 704 | ||
AK193757.1 | - | 1 | 63 | |||
BC068172.1 | - | 3 | 772 | 30550909 | ||
S55549.1 | - | 1 | 81 |