Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ubxn8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178648.2 | NP_848763.2 | 4 | 390 | ||
GenBank mRNA | AK011496.1 | - | 4 | 378 | ||
AK132565.1 | - | 4 | 383 | |||
AK161397.1 | - | 4 | 785 | |||
BC024492.1 | - | 4 | 384 | 4981720 | ||
D88447.1 | - | 4 | 373 |