Search
Search

Obfc1| Gene Symbol: Obfc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175360.2 | NP_780569.1 | 3 | 416 | ||
| GenBank mRNA | AK075574.1 | - | 3 | 416 | ||
| AK135228.1 | - | 3 | 434 | |||
| AK146929.1 | - | 3 | 494 | |||
| AK164386.1 | - | 3 | 426 | |||
| AK166348.1 | - | 3 | 490 | |||
| BC029548.1 | - | 3 | 456 | 5012064 | ||