Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sp110 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030194.1 | NP_084470.1 | 3, 4 | 510 | ||
NM_175397.4 | NP_780606.3 | 4, 5 | 536 | |||
GenBank mRNA | AK019873.1 | - | 2, 3 | 450 | ||
AK077800.1 | - | 3, 4 | 467 | |||
AK150373.1 | - | 3, 4 | 454 | |||
AY845948.1 | - | 3, 4 | 517 | |||
BC018413.1 | - | 3, 4 | 486 | |||
BC035444.1 | - | 3, 4 | 486 | |||
BC039938.1 | - | 513 | 4008284 | |||
BC094500.1 | - | 3, 4 | 478 | 4936681 | ||
BC095975.1 | - | 2, 3 | 352 |