siRNA Details
Gene Symbol: Sp110 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030194.1 | NP_084470.1 | 3 | 381 | ||
NM_175397.4 | NP_780606.3 | 4 | 407 | |||
GenBank mRNA | AK019873.1 | - | 2 | 321 | ||
AK077800.1 | - | 3 | 338 | |||
AK150373.1 | - | 3 | 325 | |||
AY845948.1 | - | 3 | 388 | |||
BC018413.1 | - | 3 | 357 | |||
BC035444.1 | - | 3 | 357 | |||
BC039938.1 | - | 384 | 4008284 | |||
BC094500.1 | - | 3 | 349 | 4936681 | ||
BC095975.1 | - | 2 | 223 |
Gene Symbol: LOC100041034 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK139759.1 | - | 4 | 465 | ||
AK163333.1 | - | 4 | 1760 |

More Information
Other Species Targeted

Panther Classification:
