Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ube2q2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_180600.3 | NP_850931.2 | 8 | 1042 | ||
GenBank mRNA | AK076148.1 | - | 8 | 905 | ||
AK083216.1 | - | 8 | 1035 | |||
AK150776.1 | - | 8 | 867 | |||
BC029111.1 | - | 8 | 871 | 2631258 |