Search
Search

P2RX1| Gene Symbol: P2RX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002558.3 | NP_002549.1 | 6 | 993 | ||
| XM_011523896.2 | XP_011522198.1 | 7 | 1385 | |||
| XM_011523897.2 | XP_011522199.1 | 6 | 833 | |||
| XM_011523899.2 | XP_011522201.1 | 6 | 1497 | |||
| XM_011523900.2 | XP_011522202.1 | 6 | 1669 | |||
| GenBank mRNA | AF020498.1 | - | 6 | 828 | ||
| AK313264.1 | - | 6 | 792 | |||
| BC027949.1 | - | 5 | 1075 | |||
| BC044657.1 | - | 6 | 768 | 5756349 | ||
| U45448.1 | - | 6 | 748 | |||
| X83688.1 | - | 6 | 725 | |||