Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bmp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173404.3 | NP_775580.1 | 2 | 1362 | ||
GenBank mRNA | AK033644.1 | - | 2 | 1000 | ||
AK035299.1 | - | 2 | 1388 | |||
AK035393.1 | - | 2 | 1351 | |||
AK037225.1 | - | 2 | 1343 | |||
AK040389.1 | - | 2 | 1363 | |||
BC117749.2 | - | 2 | 975 | 40090489 |