Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Manba | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027288.3 | NP_081564.3 | 5 | 683 | ||
GenBank mRNA | AF306557.1 | - | 5 | 687 | ||
AK036505.1 | - | 6 | 783 | |||
AK047057.1 | - | 5 | 676 | |||
AK142165.1 | - | 5 | 648 | |||
AK154172.1 | - | 5 | 664 | |||
AK166022.1 | - | 5 | 665 | |||
AK166340.1 | - | 5 | 681 | |||
AK170290.1 | - | 5 | 680 | |||
BC031409.1 | - | 5 | 626 | 4952710 |