siRNA Details
Gene Symbol: Dync2h1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029851.2 | NP_084127.2 | 60 | 9496 | ||
GenBank mRNA | AK047928.1 | - | 15 | 1875 | ||
AK173324.1 | - | 2 | 167 | |||
AY452067.1 | - | 2 | 184 | |||
DQ104402.1 | - | 60 | 9496 |

More Information
Other Species Targeted

Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Panther Classification:
