Contact Us
Four key reasons respiratory allergy diagnostics are essential in primary care
Article
Respiratory allergy
Published: February 2025
Medically reviewed by: 
Magnus Borres, Director of Medical Affairs
Think it’s allergy symptoms? It’s time to test. Did you know there is a test available to help your allergy diagnosis?
Patients presenting with symptoms like sneezing, itchy eyes, and a runny nose look to you not only for answers, but for relief.
Yet, symptoms and history alone might not be enough to know the root cause – is it the new puppy at home or the high pollen count outdoors? Could it be dust mites in their bedding or perhaps an allergy to mold or mites?
The bottom line is: specific lgE blood testing is a convenient and accurate way to identify what's causing their symptoms.
A young couple walking through the woods with "Time to Test" descorative text
Here are four reasons to put your patients' symptoms to the test in primary care:
1. Improve patients’ quality of life
While rhinitis symptoms aren't the most concerning symptoms seen in primary care, they have a significant impact on an individual's quality of life.1 If untreated, allergic rhinitis can contribute to the development of allergic asthma.2
2. Achieve diagnostic clarity for target care
Helping patients determine if their symptoms are lgE-mediated or not can aid in your diagnosis. The symptom overlap between allergic and non-allergic rhinitis makes it difficult to diagnose which disease is causing symptoms through patient history and physical exam alone.3
Think about this:
Up to 80% of allergic patients are sensitized to more than one allergen, making specific lgE testing essential for effective care.3
3. Leverage the value of a negative lgE result
There's power in a negative specific lgE blood result, too.4 A negative result suggests that additional investigation of the underlying causes of allergy-like symptoms is required. Patients with negative specific lgE blood test results may need to avoid allergens and/or take symptomatic medications, like antihistamines, that can be costly, likely ineffective and can cause side effects.3
Quick insight:
In one study, approximately 65% of patients diagnosed with allergic rhinitis and prescribed antihistamines had symptoms that were not due to allergy.5
Negative specific lgE blood test results can enable:
  • Additional investigation
  • Appropriate avoidance of allergens
  • Less money spent on antihistamines
4. Elevate primary care allergy management
Allergy management in primary care is critical.3 Primary care is often the first place a patient turns to after over-the-counter medications don't seem to work. This affords an opportunity for clinicians to help patients identify specific allergens impacting their life and wellbeing.
Take action for patients' respiratory symptoms with ImmunoCAP Specific IgE testing
When in doubt, test and take control. ImmununoCAP™ Specific lgE blood testing can help give you a comprehensive view of a patient's suspected allergies through:
1. Accurately identifying specific allergen sensitization in patients to help confirm allergies.6
2. Quantifiable results for tested allergens that can help guide rhinitis and asthma management in the context of the patient's clinical presentation.3
 3. Leveraging lmmunoCAP Specific lgE testing that is considered the gold standard for in vitro allergy diagnostics,7 supported by more than 4,000 scientific articles.8
Time to test patients with respiratory symptoms
Learn more about how respiratory allergy diagnostics can help your practice, and in turn, help enhance patient care.
References
  1. Canonica GW et al. (2008) Patient perceptions of allergic: rhinitis and quality of life: findings from a survey conducted in Europe and the United Stales. World Allergy Organ J. 1(9): 138-144.
  2. Bousquet J et al. (2008) Allergic rhinitis and its impact on asthma (ARIA) update (in collaboration with the World Health Organization, GA(2)LEN and AllerGen). Allergy. 63 Suppl 86:8-160.
  3. Demoly P et al. (2022) A pragmatic primary practice approach to using specific lgE in allergy testing in asthma diagnosis, management, and referral. J Asthma Allergy. 15:1069-1080.
  4. Thorpe M, et al. (2023) History and utility of specific lgE cutoff levels: what is the relevance for allergy diagnosis? J Allergy Clin lmmunol Pract. 11:3021-3029.
  5. Szeinbach SL et al. (2004) Identification of allergic disease among users of antihistamines. J Manag Care Pharm. 10(3):234-238.
  6. Wood RA et al. (1999) A comparison of skin prick tests, intradermal skin tests, and RASTs in the diagnosis of cat allergy. J Allergy Clin lmmunol. 103(5 Pt 1):773-9
  7. Crameri R (2013) The crux with reliable in vitro and in vivo diagnosis of allergy. AIIergy. 68(6):693-094.
  8. Ansolegui IJ et al. (2020) lgE allergy diagnostics and other relevant tests in allergy, a World Allergy Organization position paper. World Allergy Organ J. 13(2):100080.