Talk with Us
Episode 058
Early-Life Allergy Risk, Precision Diagnostics, and the Future of Prevention (ft. Dr. Anna Asarnoj)
Episode summary
A ~7% sensitization rate by 3 months of age highlights how early allergic disease can begin, making early recognition critical for long-term patient outcomes. In this episode, internationally recognized allergy researcher and pediatric allergist Dr. Anna Asarnoj shares insights from the landmark BAMSE birth cohort and her decades of research on how allergies develop, evolve, and sometimes resolve over time. Together, we explore the atopic march, early-life sensitization, component-resolved diagnostics, pet allergen risk stratification, oral immunotherapy, and emerging strategies to prevent food allergy before it becomes clinically established. Gain practical, evidence-based insights to improve diagnostic precision, risk assessment, and long-term allergy management in primary care.
Guest spotlight
Dr. Anna Asarnoj 
Dr. Anna Asarnoj is the Global Medical Director for Allergy at Thermo Fisher Scientific and a pediatric allergy and immunology physician-scientist. Formerly affiliated with the Karolinska Institute and Sachs Children and Youth Hospital in Stockholm, her research has focused on the landmark BAMSE birth cohort, allergen component diagnostics, food allergy, and the early-life factors that shape allergic disease. Her work has helped advance precision allergy diagnostics and improve risk assessment and management for patients with allergic conditions.
Episode resources
Find the resources mentioned in this episode
Lab Ordering Guide
Looking for allergy diagnostic codes from the labs you already use?
Episode transcript
Time stamps
0:00 – Intro & Welcome Dr. Asarnoj
1:32 – The BAMSI Cohort Explained
3:47 – Landmark Component Allergy Discoveries
4:16 – Birch-Peanut Cross-Reactivity
5:34 – The Peanut Challenge Study
9:11 – Early-Life Risk Stratification
11:43 – Understanding the Atopic March
12:47 – Pet Allergen Components & Prognosis
17:01 – Oral Immunotherapy in Toddlers
21:27 – Early Food Introduction & Testing
24:03 – Preventing Food Allergy
25:11 – The Future of Precision Diagnostics
26:37 – Debunking Airborne Peanut Allergy
28:38 – Eczema, Food Avoidance & Risk
30:52 – Advice for Future Allergy Researchers
32:16 – The Plasticity of the Immune System
33:37 – Closing Remarks
Transcripts
Gary (00:04):
I'm Gary Falcetano, a licensed and board certified PA with over 12 years experience in allergy and immunology.
Luke (00:12):
And I'm Luke Lemons with over six years experience writing for healthcare providers and educating on allergies. You're listening to Immunocast, your source for medically and scientifically backed allergy insights.
Gary (00:27):
Today we're especially pleased to welcome Dr. Anna Asarnoj to Immunicast. She's been affiliated with Karolinska Institute and Sachs Children and Youth Hospital in Stockholm, where she's combined clinical care with impactful research in pediatric allergy and immunology.
Luke (00:43):
And we're also proud to share that Dr. Asarnoj is now the global medical director for allergy and joined at Thermo Fisher Scientific, so a colleague as well, so not just a guest today. And we're really excited to have her on the podcast to share her experience, her global perspective and her vision for the future of allergy care.
Gary (01:04):
Yeah. Welcome Dr. Arsenai.
Anna (01:06):
Thanks a lot. And I'm also very happy to be here and to have joined you. Great team. Thank you.
Gary (01:13):
You've done so much research in the past, and I understand you're going to continue to do research even as you join us as our full-time global allergy medical director. But one of the, I think, most interesting for me anyway, areas of your research has centered around the BAMSE cohort. So can you tell us a little bit about the cohort and then what kind of research you've done with it?
Anna (01:35):
Yeah, so the BAMSE cohort, the acronym is a bit weird because it's in Swedish. So the first letter B stands for born. That is in Swedish, it means children. So it's born and children and then allergy, milieu, that is the same above the same word. And then Stockholm, the capital of Sweden, and then epidemiology, that is what it's the main aim of the study. Gotcha. And
Gary (02:03):
So this cohort, tell us a little bit about the cohort.
Anna (02:06):
It consists of about 4,000 children that were born in the 1992 and 1994, between those years in Stockholm, both city and rural areas of Stockholm. We have followed them for many, many years. So now they are soon turning 32. Wow. And
Gary (02:26):
So this is an unselected population-based?
Anna (02:29):
It's a population-based unselected birth cohort. And we have had clinical checkups four times at four, age, 16, and 24 years of age. And in between we also had questionnaire checkups, but they came to the clinic, the research clinic four times. Some of them - More.
Gary (02:49):
More than
Anna (02:50):
That. More than that.
Gary (02:51):
And you've collected numerous different
Anna (02:54):
Biomarkers
Gary (02:55):
And samples.
Anna (02:56):
And did lung functions tests and IgE testing.
Gary (03:00):
Understood.
Anna (03:01):
So I have been focused on the IgE results from this cohort, especially with the components and the longitudinal aspects of what happens during childhood.
Luke (03:13):
Can you share some of the surprising findings that you found within the cohort?
Anna (03:18):
I was really lucky to be a part of the exploding discovery of the use of component allergens or allergen components. When I started my PhD project, the aim was to... The first paper was basically just describing sensitization between four and eight years of age because that was how far we had come in the AI cohort. Right. It was that early on in the cohort.
Gary (03:42):
Yeah.
Anna (03:42):
Then we saw that the birch allergic individuals, they had so much peanut sensitization so it didn't make sense and they didn't report peanut allergy in that extent. So we were a bit confused about that, but then we started to look at the components and the Ara h 8 versus ARA h 1, 2, and 3 sensitization.
Gary (04:08):
That was some of the early work then that really helped identify bird sensitization driving what we call asymptomatic peanut sensitization.
Anna (04:17):
Exactly. So I can actually, I'm quite proud of making that contribution to science that we had this hypothesis that if you're only sensitized to Ara h 8, you're actually basically birch allergic or polynallergic. So you might not have to worry about your peanut sensitization because they were also sensitized to peanut extract. Exactly. So then when we looked at the children that had negative IgE to Ara h 1, two, and three, the storage proteins that we now know is connected with genuine peanut allergy. So children that were negative in those components, but positive for RIH8, we had them have a peanut challenge.
Gary (05:03):
And that was really one of the first studies to look at broadly monosensitized Ara h 8 patients.
Anna (05:11):
We were a bit surprised when the first patient we had in the clinic, he had a massive euanaphylaxis.
Gary (05:19):
No, this is something we were talking about the other day. It was the first patient entered into the study. I see, I never knew that. Oh my gosh.
Anna (05:28):
No, that was really... I mean, we didn't know. I mean, we were shocked.
Gary (05:33):
Yeah.
Anna (05:33):
I mean,
Gary (05:34):
Because here your hypothesis is that Ara h 8 is relatively benign
Anna (05:39):
And doesn't usually
Gary (05:40):
Cause
Anna (05:40):
Symptoms. No, no. But at that times we couldn't measure or we could, but we had in our material we had only a sensitization above 0.35 kilometers per liter. So it turned out that this boy had IG to Ara h 2. I think it was 0.22, but he had to Ara h 6, like a high level of 20 something.
Gary (06:05):
So I do want to take a second and just refer our listeners back. We have done previous episodes on both tree nuts and peanuts where we talk about this in detail. But I think you really in that study started to really clarify that it isn't just Ara h 2 or one in three for that matter, that Ara h 6, which is a very similar protein to Ara h 2 can be very significant and that is why it has been added into the available peanut allergy components. So
Anna (06:35):
Actually we wrote the case report of that first patient because the rest of the, I don't remember exact numbers, but it was many, many children coming in for challenges and none of the others reacted. So our hypothesis was correct.
Gary (06:52):
But with this exception - I can tell you, because I speak about this study so often, it was a total of 144 patients I remember. So for you, it's kind of distant memory, but it's such landmark information that I do speak about it in educations on a fairly regular basis.
Anna (07:11):
That's great.
Gary (07:11):
Yeah.
Anna (07:12):
No, so that case report with the boy that had an anaphylaxis because of his aura H6 sensitization, that was very interesting to get that knowledge maybe in an unnecessarily hard way for me as a young PhD student. Right.
Gary (07:29):
But he did do fine. You
Anna (07:31):
Responded. Yeah, of course. But he needed to be taken care of in the intensive care unit. So I mean it was serious. Very
Gary (07:39):
Significant.
Anna (07:40):
Yeah. But that case is also very well-cited case report. In the end, I'm happy I gained knowledge and others too.
Gary (07:49):
Exactly. And we're able to share it.
Luke (07:50):
And there's a lot of great stuff within the cohort. And I'm curious for primary care providers out there who maybe see infants with eczema or early wheeze based on some of the data in the BAMC cohort, how should they think about risk stratifying those patients?
Anna (08:06):
What we see throughout all of our studies is that it's what happens early in life that is really impacting the rest of the live. So for example, if we stay with peanut allergy for a while, we took the first blood samples at four years of age. I wish we had some earlier samples, but we don't. But I have in another study that we can talk about, but in this study we had the first blood samples at four. And actually about peanut allergy, not much happens with storage protein sensitization. It's already set at four years of age. So it's only a few individuals that will get rid of their sensitization or get the novel sensitization up to the age of 24. And I guess it'll also continue further. That's really interesting that the food allergies are early in life.
Gary (09:04):
That really speaks to our new knowledge, relatively new in the last decade of early introduction and the dual exposure hypothesis of getting those allergens into the diet as soon as possible to help prevent development. So true. Yeah.
Anna (09:19):
And then we looked at in another cohort, preventative cohort that I also have a few papers from, that already at three months of age you have sensitization in 7% of the children.
Gary (09:32):
Was there a relationship there to preexisting skin disease, atopic dermatitis or not?
Anna (09:38):
Yeah, you can also see early dry skin, early eczema. Everything is connected to allergic development of allergic diseases and sensitization. So it's all the atopic march.
Gary (09:53):
Yes, right. Interrelated. Yeah. And I think that's exactly what Luke was referring to.
Luke (09:57):
Yeah, yeah. Just for the primary care providers out there as well, it's incredible research that helps us risk stratify those early on patients. And like you said, the atopic march, just to make sure that everyone is growing up healthy. It's very important to catch those symptoms in patients when they're younger so that we can prepare later on in life.
Anna (10:20):
Yeah, exactly. So what you mean is with the atopic march is that you begin with early in life, you have eczema and food allergy and then during childhood you will get asthma and eventually rhinoconjunctivitis. So it's like a trajectory that you are at high risk of follow during childhood. Yes, exactly.
Gary (10:40):
Exactly.
Anna (10:41):
And not
Gary (10:41):
Every patient follows that
Anna (10:42):
Trajectory,
Gary (10:43):
Right?
Anna (10:43):
But that
Gary (10:43):
Is a very common
Anna (10:44):
Trajectory. So one very, very strong risk factor is early sensitization. So if you have asthma, for example, if you have a toddler with asthma, if the toddler is not sensitized, he or she would probably grow out of this asthma and not have difficulties with allergies or allergic disease. But if you're sensitized, you're at much higher risk.
Gary (11:09):
So speaking of that, getting back to components and outside of food components, you also did a lot of work around pet allergen components and had some insights from the BAMC cohort as well.
Anna (11:24):
So we looked at cat and dog allergen components and saw a very strong relationship between being sensitized to more numbers of components. You are at higher risk of having the allergy.
Gary (11:40):
And even maybe a more severe phenotype of the allergy, right?
Anna (11:44):
Yeah, more severe phenotype as well. So it's really a strong, strong prognostic marker and much stronger association than, for example, just looking at the extract sensitization. So it's a difference.
Gary (12:01):
And there was also almost a predictive aspect as well in the four-year-old cohort to when they were 16.
Anna (12:08):
Yes.
Gary (12:08):
What did you see there that was interesting?
Anna (12:11):
Yeah, we saw the same thing that the more components you had early sensitization to, the more likely that you ended up with an allergy.
Gary (12:20):
Right. Or additional respiratory disease from what I remember, correct? If they only had a mild disease at age four, they may have asthma at
Anna (12:27):
Age 16. And we also see that we can see that for cat and dog allergy, but we have another publication when we also see that at four years of age, a few allergens, if you're sensitized to them very, very predictive for future asthma or rhinoconjunctivitis at 16 years of age. Now we're looking also at the endpoint 24 years of age.
Luke (12:55):
For the primary care audience out there, what does that mean if they have a child come in, let's say, who is sensitized to one of those pet allergen components and is already experiencing maybe some respiratory issues? Are you saying that that potentially should be monitored and evaluated for their asthma going forward? Or what would you recommend for primary care providers?
Anna (13:18):
Yeah. So if I had a child or if I have a child in the clinic that is multisensitized, I will be much more active in the treatment and also looking at, I mean, maybe there is a cat or dog in the home, then I might just plant the idea of having a plan B for that pet ownership.
Luke (13:42):
But there's also, I think we mention it often here around the pet allergen component Can f 5, because if it is a male dog, they may be able to tolerate a female dog. So Can f 5 is only produced in the prostate of the male dog. And there's a lot we have on pet allergen components and I recommend checking out that episode if you're listening now because it is a difficult conversation and the work that you've done to show how important pet allergen components are for the patient is incredible too.
Gary (14:12):
Yeah. I've seen various reports of monosensitization to Can f 5 in different studies. What did your experience show? So again, people that are just sensitized to Can f 5.
Anna (14:22):
In the BAMS study that we talk a lot about, we saw that monosensitization to Can f 5 was very common and many of those actually didn't report any symptoms to dog. So unfortunately, we don't know which gender dogs they were supposed to, but it wasn't as strong relationships as with, for example, Can f 1 that has the strongest relationship to symptoms
Gary (14:48):
In obviously both male and female dogs. Yeah.
Luke (14:53):
So pivoting a little bit into a different field, oral immunotherapy, you are currently involved in some research with that, and I'm curious if you can share some information with our listeners today.
Anna (15:06):
We know from other studies that oral immunotherapy for food allergy isn't working that good in adults. The effect is so- so and there are lots of side effects. You can see that there might be an age gradient so that the younger you are, the better the therapy works. So we have just finished a study with toddlers one to three years of age when they start with peanut allergy that we randomized either to have a oral immunotherapy and we used the peanut puffs, Bambas. The Bambas, yeah. Or continuing to avoid peanuts. So it was not blinded, but it's difficult to have a blinded placebo puff. Anyway, we just finished that study and it was really, really good results. I mean most of the children actually like to eat the Bambas. They're
Gary (16:04):
Pretty good, I have to say. Yeah. Yeah, they
Anna (16:06):
Taste good.
Luke (16:07):
Part of the study.
Anna (16:09):
So the compliance and feasibility was really good. And we also had quite long up-dosing intervals. So with oral immunotherapy, you start eating a very, very low dose of the allergen and then you increase the dose. And so we used four to six weeks between the increased steps and that also turned out to be really good. It was actually quite many of the participants were recruited during the COVID pandemic. So it was good because they were not so sick in other infections. But when the COVID ends, they got very, very sick. Yeah. Everywhere. Yeah. So there was a lot of, they had planned visits for dose escalation, but they got sick. And then it was really good that we had these quite long intervals. Yeah, like a
Gary (17:03):
Real world.
Anna (17:04):
Yeah.
Gary (17:04):
It
Anna (17:05):
Was really a good protocol that we had. And the effect was great and we had very few side effects. So we think we are so satisfied with
Gary (17:15):
Resources. That's very encouraging. And I think for our US colleagues, especially our US primary care colleagues, we've only had one FDA cleared oral immunotherapy, Palforzia, which I think is being discontinued. Yes, it is. But there are a lot of oral immunotherapy programs around the country. I think it's important for our primary care colleagues to find local allergists that are doing and get connected with them if patients are interested.
Anna (17:43):
And now we got so inspired by our experiences and the families were so happy. But to be honest, some of the families were also a bit annoyed because they said to us, "Okay, we are so grateful for this with the peanut allergy treatment, but what about the egg allergy or the
Gary (18:03):
Hazelnut
Anna (18:04):
Allergy?"
Gary (18:05):
Because most of those patients have
Anna (18:07):
Multiple allergies, right? Multiple treatment to that. So now we have actually started a new oral immunotherapy study with multi-allergen. So they get the powder containing 10 different foods and then they eat that in increasing doses. We'll be anxious to hear
Gary (18:27):
How that
Anna (18:27):
Goes. It's very exciting to follow what happens. And
Luke (18:31):
I just realized this somewhat connects to what we were talking about earlier when it comes to children early on in the atopic March and around testing before exposing them to certain types of foods like the peanut puffs.
Anna (18:45):
I'll
Luke (18:45):
Go back again to that and ask, what should primary care providers know related to food allergy management for young children? Exactly.
Anna (18:53):
So that's a really, really good question because I mean, we have been advising that you should only test for foods that you have a clinical history with a suspicion of allergy because there is a situation with false positive IgE results for foods that can make families make decisions of an unnecessary avoidance of foods. But if in the future, if we think about this with early introduction, maybe we need some more testing to see which foods that are really important to introduce. Because if you can see that you have a sensitization coming up for a certain food, then it's really important that you treat that.
Gary (19:37):
So if you have an early, especially maybe a low level sensitization to a food but can tolerate it, that may be the most important food to get
Anna (19:44):
Into the
Gary (19:45):
Diarrhea.
Anna (19:45):
You have a window of opportunity to do something about it before it becomes clinical.
Gary (19:51):
This
Anna (19:51):
Is just a hypothesis. But I'm just speculating for the future. It might be a way of handling this in the future. Exactly.
Gary (20:01):
And then for patients that are at high risk and the parents and the caregivers are really reluctant to do early introduction, that's another place where this strategic use of testing can help to reassure with the negative to get them to introduce -
Anna (20:18):
That's also a use that we cannot test for all foods in the whole world, but in some situations it's the most feasible way to do it to just show that, look, there is no- No, so
Gary (20:33):
Please start feeding.
Anna (20:35):
But I mean, it's difficult because then you don't want parents coming in then every week and want another test before they - Exactly, exactly. I mean it's good with the testing, but it's not ideal. No,
Gary (20:48):
That's not ideal.
Luke (20:49):
Yeah. So you had mentioned a hypothesis. And so out of curiosity, what are some of the questions or unanswered questions and allergy right now that excite you the most that you want to know more about? You had mentioned this hypothesis around food. Is there any other sort of subject that you want to know more about?
Anna (21:11):
Exactly. So now I'm quite occupied by this thought of that you have the window of opportunity to do something about an uprising allergy or I mean better to prevent the allergy from the beginning with the early introduction before one year of age that we now know it is recommended. So yes, to find a way to stop the increase of food allergies is quite in my research now. But I have of course more ideas if I think.
Luke (21:44):
Yeah, no I mean there's so much, but it is really interesting if we could figure out that window,
Anna (21:50):
The
Luke (21:50):
Amount of help that we could provide these children would be great. I
Anna (21:53):
Think it's like a paradigm shift now coming up. Absolutely. Yeah,
Gary (21:59):
I hope so.
Anna (22:00):
It's really exciting and it's fun to be a part of it. Yeah,
Luke (22:04):
To look forward into the future. And speaking of, where do you see precision diagnostics existing in that space? Yeah,
Anna (22:11):
So I think there is a very important role for diagnostics as I already mentioned because we might shift the way we think of it because of course we want to avoid unnecessary avoidance because of false positive testing, but the positive test can also be a signal that we have to do something.
Gary (22:33):
Yeah. And that actually speaks to the benefit, especially for the foods that we have allergen components, because it does help to classify or qualify those positives as to being more significant or less significant as we talked about with the peanut example.
Anna (22:50):
And also for egg. Yes. The ovomucoid, the IgE test is really a good prognostic value of that.
Gary (22:59):
Exactly. So not just the ability to tolerate a baked form, but also we've seen with ovomucoid and with casein with milk, those are more prognostic of maybe continuing.
Anna (23:12):
That's also a potential. So it's very exciting.
Luke (23:16):
And what is one misconception about allergy that you wish would disappear? Let's say you snap your fingers and just go away. Everybody would forget about the misconception.
Anna (23:27):
Well, it may be a bit off-topic, but the misconception of airborne peanut allergy.
Gary (23:33):
Oh, yeah. I mean the plain anxieties and -
Anna (23:37):
Yeah, unnecessary anxiety. I try to tell my peanut allergic patients that they don't have to worry if they feel the smell of peanuts. It's a very unpleasant smell if you're allergic. I like the smell, but I can understand that if you are peanut allergic that you make an association that this is dangerous. But to try to comfort them that the proteins that triggers the reaction that can be dangerous, they are not flying around. You have to eat the protein to get a reaction.
Gary (24:10):
I think that's so important because there have been reports and I think the literature shows that anxiety can almost be a precipitating factor in a mast cell release, right?
Anna (24:23):
Yeah.
Gary (24:23):
So you think you're going to react, so you actually do show some symptoms,
Anna (24:28):
Right? Yeah, but you can never have severe symptoms from the fear that is important to point out. So there is actually no deaths reported ever from airborne exposure to peanut. There aren't any ever.
Gary (24:46):
Well, I think that's a great topic then. I think that's a great thing to put to rest.
Luke (24:50):
I think it's also important for our listeners to hear that as well. If they do have patients who come in and have a peanut allergy and they express that where we've talked to some mental health allergy therapists before and the anxiety that follows a lot of these
Anna (25:04):
Patients is
Luke (25:05):
Crippling sometimes.
Anna (25:06):
Yes, it really is. That is one misconception that I would like to. Okay. Yeah. But I also have one. Yeah, what is it? And then that is a very common situation when you have parents seeking advice for their baby with eczema and they are seeking for the cause of the eczema. And the cause, the suspicious from the parent is quite often that it's some food. Yes. Yeah. And if you then in that situation start to avoid foods, then you actually will increase the risk for that child to develop allergy to that food. I'm
Gary (25:46):
So happy that you brought that up.
Anna (25:48):
Yeah, because that is so common. And I understand that you want to not risk anything, but in fact, you are putting your baby at risk when you do that.
Gary (25:58):
Exactly. And I would definitely refer our listeners to a previous recent episode on atopic dermatitis that we did where we discussed this specific topic and how the paradigm has really changed from it being eczema or atopic dermatitis being driven by a food allergy to just the opposite that the skin barrier issue actually sets you up for future food allergy. And I think as a discipline, allergy is really starting to understand that well now. Yeah.
Anna (26:28):
But you also have to educate the parents.
Gary (26:31):
Yes, and primary care providers.
Anna (26:33):
And primary care providers, definitely, but the parents are also making some decisions on their own without even consulting anyone. So I meet more and more families that are aware.
Gary (26:47):
That's
Anna (26:47):
Good. That's great. We have to make
Gary (26:49):
Sure we get the right information out there. I
Anna (26:50):
Think the trend is in the right direction.
Gary (26:53):
So one question before we finish, I know you have really married your clinical career with a research career almost from the beginning. Is that correct?
Anna (27:02):
Yeah, almost from the beginning. Yeah.
Gary (27:04):
Yeah. What advice would you have for young clinicians or researchers who are interested in allergy research and are considering doing something similar?
Anna (27:13):
I would really recommend it.
(27:16):
I mean, it's such a good mix to whatever you do. I would always recommend to do something that is not the same thing every day, that you do different things and learn from different areas. And it's much more fun to go to work. And it's also make the years when you have young kids and because with the research you can a little bit more decide yourself how you use your time. And the clinic is the clinic. The patients are booked. Exactly. And that mix is very good to have at some week you can plan yourself and the other week you're on the schedule. Yeah, on the schedule. So it's a really good mix. Yeah, I think that's great
Gary (28:00):
Advice.
Anna (28:01):
Yeah.
Luke (28:02):
Just diversity of research, diversity of the way you work.
Anna (28:05):
Yes.
Luke (28:06):
Again, looking at all your research, you've definitely embodied that diversity and helped push us further in the field of allergy. And I
Gary (28:12):
Can't wait to see
Luke (28:13):
Where we
Gary (28:14):
Go in the future.
Luke (28:15):
Exactly. Yeah. On a personal note, after decades of studying allergic diseases, what is one thing that still fascinates you about the immune system?
Anna (28:24):
Interesting question. I guess now I'm thinking a lot of the flexibility of the immune system. That is, during childhood, I think my first publication was named The Dynamic Process of something. That is really true. It's so dynamic and it is the immune system that is changing. And especially with a food allergy that is the first years, the plasticity of the immune system that seems to end at some point. It's really fascinating.
Gary (29:00):
Yeah, that is interesting.
Anna (29:01):
And that it's not true for inhalant allergen. I mean, you get sensitized later on. You have also early sensitization, but most of those sensitization is actually a bit later. Right.
Gary (29:13):
And there doesn't seem a way of interrupting that progression with
Anna (29:17):
Inhalants, right? Yeah, it's fascinating. Yeah, the
Luke (29:20):
Dynamics of it all. Yeah. Well, again, thank you so much for coming onto ImmunoCast.
Anna (29:26):
Thank you. And thank you for having me.
Gary (29:29):
Yeah, and thank you for joining us as a
Anna (29:31):
Colleague as well. Yeah, so happy.
Gary (29:32):
We're very excited to
Anna (29:33):
Have you. Yeah, me too.
Luke (29:34):
And for everyone out there listening, in the description of this episode, you'll find a link to this episode's webpage where we'll have resources and links and some additional episodes that are around some of the topics that we've talked today.
Gary (29:47):
Yeah. Thanks again for listening and we'll see you next time.
Speaker 4 (29:56):
Immunocast is brought to you by Thermo Fisher Scientific, creators of ImmunoCAP Specific IgE diagnostics and Phadia laboratory systems. For more information on allergies and specific IgE testing, please visit thermofisher.com/immunocast. Specific IgE testing is an aid to healthcare providers in the diagnosis of allergy and cannot alone diagnose a clinical allergy. Clinical history alongside specific IgE testing is needed to diagnose a clinical allergy. The content of this podcast is not intended to be and should not be interpreted as or substitute professional medical advice, diagnosis, or treatment. Any medical questions pertaining to one's own health should be discussed with a healthcare provider. This podcast was edited by Resonate Recordings. 
References used in this episode
  • Platts-Mills TA, Tovey ER, Mitchell EB, Moszoro H, Nock P, Wilkins SR. Reduction of bronchial hyperreactivity during prolonged allergen avoidance. Lancet. 1982 Sep 25;2(8300):675-8.
  • Platts-Mills TA, Hayden ML, Chapman MD, Wilkins SR. Seasonal variation in dust mite and grass-pollen allergens in dust from the houses of patients with asthma. J Allergy Clin Immunol. 1987 May;79(5):781-91. doi: 10.1016/0091-6749(87)90211-9. PMID: 3571770.
  • Gelber LE, Seltzer LH, Bouzoukis JK, Pollart SM, Chapman MD, Platts-Mills TA. Sensitization and exposure to indoor allergens as risk factors for asthma among patients presenting to hospital. Am Rev Respir Dis. 1993 Mar;147(3):573-8. doi: 10.1164/ajrccm/147.3.573. PMID: 8442589.
  • Tovey ER, Chapman MD, Platts-Mills TA. Mite faeces are a major source of house dust allergens. Nature. 1981 Feb 12;289(5798):592-3. doi: 10.1038/289592a0. PMID: 7464922.
  • Chung, Christine H., et al. "Cetuximab-Induced Anaphylaxis and IgE Specific for Galactose-α-1,3-Galactose." The New England Journal of Medicine, vol. 358, no. 11, 2008, pp. 1109-1117. NEJM,