Contact Us
Indoor allergy in primary care: A practical approach to ImmunoCAP Specific IgE testing
Article
Respiratory allergy
Pet allergy
Published: October 2026
Medically reviewed by: 
Gary Falcetano, PA-C, AE-C
When respiratory symptoms persist, look beyond the pollen calendar
House dust mites, furry animals, mold and other indoor allergens can contribute to respiratory symptoms year-round. Combined with the patient’s history, ImmunoCAP™ Specific IgE testing can help identify sensitization, support diagnosis and inform patient management.1-3
Recognizing the patterns associated with indoor allergy can help determine when further evaluation may be appropriate.
Senior Man Blowing His Nose While Sitting at Home
When to suspect indoor allergy
Consider indoor allergy in patients with persistent or recurrent respiratory symptoms, including rhinitis or wheezing, particularly when symptoms appear to change with the patient’s environment or exposure.4 Symptoms alone cannot distinguish allergic from nonallergic causes, so look for patterns that may point to an allergic cause.
Look for patterns such as:
  • Symptoms that worsen at home or in particular indoor environments
  • Flares associated with cats, dogs or other furry animals
  • Symptoms associated with damp or mold-prone environments
  • Possible exposure to house dust mites or cockroach
  • Improvement away from a particular environment and recurrence after returning
  • Respiratory symptoms that continue outside the expected pollen season
Patient history can point to indoor allergy, but the source of exposure may not always be obvious. Common indoor allergens can be encountered in many everyday environments, and patients may be exposed to more than one.
Common indoor respiratory allergens
Common indoor allergen sources include house dust mites, furry animals, mold and cockroach.
Dustmites
House dust mites
House dust mites are commonly found indoors, particularly in bedding, upholstered furniture and other areas where dust can accumulate.
dog and cat on owner's lap
Cats and dogs
Exposure to furry animal allergens is not limited to pet owners. Allergens can be encountered in homes and other environments where cats or dogs are or have been present.
moldy wall
Mold
Indoor mold exposure is often associated with damp or moisture-prone environments. Exposure can vary depending on building conditions and local climate.
cockroach on kitchen counter
Cockroach
Cockroach allergen exposure can occur in some indoor environments, with patterns influenced by geography, housing conditions and other local factors.
When indoor allergy is suspected, start with a profile
Choosing which individual allergens to test can be challenging. Where available, a locally relevant, laboratory-defined respiratory allergen profile offers a practical first-line approach by assessing sensitization to multiple respiratory allergens at once.
Profiles using ImmunoCAPTM Specific IgE tests  may include perennial indoor allergens alongside seasonal allergens relevant to the local environment. This structured approach provides a clear starting point while reducing the need to select allergens one by one.2
Follow your laboratory’s local profile name, code and ordering pathway.
From sensitization to clinical relevance
Once results are available, the next step is to determine which sensitizations are clinically relevant. A positive specific IgE result indicates sensitization but does not, by itself, establish clinical allergy.3  If the clinical picture remains complex, additional evaluation or specialist referral may be appropriate.
Positive results
Consider whether the result aligns with the patient’s symptoms and exposure history. For example, house dust mite sensitization may be more clinically relevant when persistent respiratory symptoms are associated with the home environment.
badge icon
Negative results
If sensitization to a suspected indoor allergen is not detected, consider whether another allergen or a nonallergic cause may better explain the patient’s symptoms.
people icon
Multiple positive results
When more than one sensitization is identified, consider each finding alongside the patient’s symptoms and exposures. For example, positive results to both house dust mite and cat may reflect different exposure patterns, with the clinical history helping determine which findings are relevant.
Learn more about how to interpret results. 
From uncertainty to targeted care
If clinically relevant sensitizations are identified, ImmunoCAP Specific IgE test results can support a more targeted approach to patient care.
For patients with indoor allergy, results can help inform:2-4
  • Targeted management, including appropriate treatment and allergen avoidance or exposure-reduction strategies
  • Clear patient conversations about allergens that may be contributing to symptoms
  • Appropriate referral when symptoms, multiple sensitizations or treatment needs warrant specialist evaluation
Frequently asked questions
Yes. Indoor allergens such as house dust mites, furry animals and mold can remain relevant outside a defined pollen season. Indoor and seasonal exposures may also overlap.
Yes. Patients can be sensitized to multiple allergens, and indoor and seasonal exposures may overlap. Consider each result alongside the patient’s symptoms, exposure history and overall clinical picture.
Interpret the results in the full clinical context. Revisit the patient’s symptoms and exposure history and consider whether another allergen or cause better explains the presentation. Additional evaluation or specialist referral may be appropriate when uncertainty remains.
Yes. If the initial results do not fully explain the patient’s symptoms, additional specific IgE testing or, where available, testing to allergen components may provide more detail. The appropriate next step depends on the clinical picture and local testing pathways.
ImmunoCAP Specific IgE profile is not a CE-marked product and is not placed on the market by Thermo Fisher Scientific. It is a laboratory-defined testing panel, consisting of individually CE-marked ImmunoCAP™ allergen products ordered separately by their official product names and article numbers.
References
  1. Wise SK, et al. Int Forum Allergy Rhinol. 2023;13(4):293-859.
  2. Demoly P, et al. J Asthma Allergy. 2022;15:1069-1080.
  3. Santos AF, et al. Allergy. 2023;78(12):3057-3076.
  4. Papadopoulos NG, et al. Allergy. 2015;70(5):474-494.