Recent searches Clear History
Contacto
Allergen Encyclopedia
Table of Contents
Componente
e231 Fel d 7
e231 Fel d 7 Scientific Information
Name; WHO/IUIS:
Fel d 7
Biological function:
Lipocalin
Allergen code:
e231
Molecular Weight:
17.5 kDa
Source Material:
Recombinant protein
Other Names :
Cat lipocalin
Summary
Fel d 7 is a lipocalin present in cat saliva, lacrymal fluid, and on cat hair. Fel d 7 sensitization is found in 31-54% of cat-allergic patients, mainly in association with secretoglobin Fel d 1. Fel d 7 belongs to a subgroup of lipocalins with clinically relevant cross-reactivity with Can f 1 from dog. Fel d 7 sensitization by itself and within complex patterns of sensitization to lipocalins is associated with persistent type 2 inflammation, and an increased risk of asthma, allergic rhinitis, and atopic dermatitis.
Epidemiology
 Worldwide distribution
Cat allergens are among the most important sensitizers and causes of IgE-mediated allergic disease worldwide, due to high rates of cat ownership, frequent indoor presence of cats, and the demonstrated dissemination of cat allergens in schools and in homes without cats [1]. Sensitization rates to cat extracts are high in both asymptomatic and symptomatic individuals [1].
The prevalence of sensitization to cat extract and allergens increases during childhood, reaching a peak in teenagers and young adults, then slowly decreases in adults and elderly individuals [2, 3].
Fel d 7 sensitization is most often found in association with other mammalian allergens, notably with the cat marker allergen Fel d 1 and the lipocalin Fel d 4 in cat-allergic patients, and with Can f 1 without Fel d 1 in dog-allergic patients [4, 5].
The prevalence of Fel d 7 sensitization varies as a function of exposure to cat allergens, atopic status, asthma or allergic rhinitis (AR), ethnicity, cohort selection, and geography [1, 2, 6].
Fel d 7 sensitization was initially reported in 37% of cat-allergic patients, although typically with low IgE levels, and in 8% of cat-tolerant individuals [7].
Later studies confirmed that Fel d 7 sensitization can be found in both symptomatic and asymptomatic individuals.
In adult European patients with demonstrated cat allergy, the prevalence of Fel d 7 sensitization was 31% - 54% [3, 8, 9]. Similar findings were reported in a Korean cohort of 238 adults, with Fel d 7 sensitization found in 67/206 (33%) of cat extract-sensitized individuals [10].
In an Austrian cohort comprising both children and adults, the prevalence of Fel d 7 sensitization was low in cat-monosensitized patients, at 3.8%, but increased to 29% in cat and dog double sensitized patients, and to 59% in cat, horse, and dog polysensitized patients [4].
The prevalence of Fel d 7 sensitization was 47% in Swedish asthmatic patients aged 10-35 years with demonstrated cat, dog or horse sensitization and requiring inhaled corticosteroid or antileukotriene treatment [11].
In a Swedish cohort of children aged 10 – 17 years investigated for furry animal allergy or sensitization, Fel d 7 sensitization was found in 68% of those experiencing cat-induced respiratory symptoms, but also in 55% of those who did not display such symptoms [6].
Among Lithuanian patients referred to outpatient allergy clinics for allergen multiplex investigations, the reported prevalence of Fel d 7 sensitization was 37% [12].
Apparent monosensitization to Fel d 7 is infrequent in genuine cat-sensitized individuals, usually attributed to cross-reactivity with dog lipocalin Can f 1, explaining the wide variations reported in the literature, from 2% to 20% [7, 9-11].
Environmental characteristics
Source and tissue
Fel d 7, a homologue of von Ebner gland proteins described in various mammals, is a secreted protein first identified from a cat tongue DNA library containing the lingual salivary glands [7]. Despite no Fel d 7 expression being evidenced in the princeps study from other salivary, skin, or anal cDNA libraries, Fel d 7 allergen was present in cat saliva and cat hair, while it was lacking from cat pelt [7]. The estimated contribution of Fel d 7 to total protein content of cat saliva and cat hair was less than 0.5% [7]. Fel d 7 was also demonstrated in the cat tear film proteome [13].
Like other lipocalins, Fel d 7 is readily conveyed by direct contact with cats and through airborne particles, explaining their presence in homes and public places.
Risk factors
Cat ownership, exposure to cats, and atopy are associated with an increased risk of becoming sensitized to cat allergens, including Fel d 7 [1]. While Fel d 1 is ubiquitous and can act as a primary sensitizer even in individuals who do not own a cat, cat ownership may be a risk factor for primary Fel d 7 sensitization [4, 9]. In dog-sensitized patients, dog ownership was not associated with an increased risk of Can f 1 – Fel d 7 cross-reactivity [4].
Clinical relevance
Specific molecules
Fel d 7, a lipocalin showing cross-reactivity with Can f 1 from dog, indicates sensitization to furry animals, but not specifically to cat [4, 5].
Cross-reactive molecules
Fel d 7 belongs to a subgroup of lipocalins which also contains Can f 1, with established clinically relevant cross-reactivity [5].
Disease severity
As reviewed in the “Worldwide distribution” section, an increased prevalence of Fel d 7 sensitization, especially in association with other cat and/or dog allergens, is reported in symptomatic cat and/or dog allergic patients, compared to asymptomatic sensitization. In addition, polysensitization to multiple lipocalins, including Fel d 7, is associated with an increased risk of symptom severity. The ability of Fel d 7 to induce IgE-dependent degranulation was demonstrated in basophil activation tests, with higher potency than other cat allergens [6, 14], while its association with persistent type 2 inflammation and more severe respiratory symptoms was addressed in a Swedish cohort of children and young adults [11].
Allergic asthma and allergic rhinitis
An early cross-sectional study in 269 children reported that asthma was associated with sensitization to the lipocalin protein family [15].
In a cohort of 266 Swedish children with well-characterized asthma and sensitization to furry animals, detectable IgE to Fel d 7 and/or Fel d 4 in addition to Fel d 1 were associated with a significantly increased number of recent asthma attacks and overall worse asthma control [11]. In the same study, Fel d 7 + Fel d 4 cosensitization was independently associated with an increased fraction of exhaled nitric oxide (FeNO) and with increased concentrations of circulating total IgE [11]. These biomarkers point to persistent type 2 inflammation, which is a major contributor to atopic diseases, in conjunction with Fel d 7 sensitization.
Current asthma and current allergic rhinitis were also associated with lipocalin (Fel d 7 and Fel d 4) sensitization in Swedish adults aged 16-75 years [9]. In this study, concomitant Fel d 1 and Fel d 7 sensitization was the most frequent pattern in individuals with allergic rhinitis (32%).
Finally, a study investigating the efficacy of subcutaneous immunotherapy reported that 42% of patients with a history of rhinitis/asthma tested positive to recombinant Fel d 7 in skin prick tests [16].
Atopic Dermatitis 
In a Russian cohort of cat-allergic adults, AD symptoms were associated with Fel d 7 sensitization in over 50% of patients [8].
Molecular aspects                  
Biochemistry
Fel d 7 is a 17.5 kDa protein belonging to the lipocalin family of animal allergens, capable of efficient airborne dissemination and comprising most mammalian allergens [5]. Lipocalins display an internal cavity for binding hydrophobic ligands, which are pheromones or odorants in the case of mammalian lipocalins [5].
Isoforms, epitopes, antibodies
As of April 30, 2024, a unique isoallergen, Fel d 7.0101, has been included in the World Health Organization (WHO) and International Union of Immunological Societies (IUIS) Allergen Nomenclature [17].
Cross-reactivity due to structural similarity
Lipocalins as a family are characterized by low sequence identity (20-30%) but conserved 3-dimensional conformation, which allows selective cross-reactivity [5]. Clinically relevant cross-reactivity between Fel d 7 and Can f 1 is supported by high sequence identity (63%) in addition to the conserved 3-dimensional conformation [5]. IgE binding inhibition studies showed that Fel d 7 cross-reacts with dog dander extracts [7].
IgE binding is highly overlapping between Fel d 7 and Can f 1 [7, 18]. However, Fel d 7 and Can f 1 also contain epitopes that are not shared, resulting in non cross-reactive IgE antibodies which bind only Fel d 7 or Can f 1 and can co-exist with Fel d 7 – Can f 1 cross-reactive IgE in the same patient [18].
Diagnostic relevance              
Disease Severity
In line with the literature review presented in the above section of “Clinical Relevance”, Fel d 7 sensitization has been associated with respiratory symptoms and disease severity on its own, in association with Fel d 1 sensitization, and within a phenotype of lipocalin or wider polysensitization [1, 2, 4, 5, 8-11].
At the clinical laboratory level, the demonstration of Fel d 7 sensitization is more frequent and displays higher IgE levels in individuals experiencing cat-related allergic symptoms, in patients with asthma, AR, AD, as well as in individuals from the general population displaying symptoms such as rhinoconjunctivitis and wheeze [1, 4, 9].
Serum concentrations of Fel d 7-specific IgE were significantly higher in asthmatic patients with higher FeNO and higher blood eosinophil counts, exhibiting a positive association with these inflammatory biomarkers, as well as with airway hyperresponsiveness and a trend to decreased lung function [11].
Cross- Reactivity
Fel d 1 cat-related clusters of sensitization, including Fel d 4, Fel d 7 and Fel d 2, are considered to be driven by Fel d 1 and correspond to genuine sensitization to cat [6]. In contrast, Fel d 7 cross-reactivity with Can f 1 is illustrated in cases of primary sensitization to dog, when Fel d 1 cosensitization is lacking [5]. Comparison of the concentrations of serum specific IgE to Fel d 7 and to Can f 1 may help identify the primary sensitizer, since virtually all patients with higher levels of IgE to Fel d 7 were cat owners in an Austrian cohort, while most of those with higher levels of IgE to Can f 1 were dog owners [4]. Overall, the cross-reactivity between Fel d 7 and Can f 1 explains why Can f 1 cannot be used as a marker allergen for dog sensitization in case of concomitant cat sensitization [5].
Importantly, irrespective of the primary sensitizer, Fel d 7 sensitization is associated to an increased risk of cat and dog-induced symptoms [1, 14].
AIT Prescription
Lipocalins including Fel d 7 are not currently considered as adequate options for molecular therapeutic approaches, due to their cross-reactivity [5, 19].
Explained results                   
Allergen information
Fel d 7 is a minor cat allergen belonging to the lipocalin family, secreted in cat saliva, spread through grooming on the fur, and readily disseminated indoors. Fel d 7 sensitization is associated with persistent type 2 inflammation.
Clinical relevance
Fel d 7 sensitization, both on its own and within complex sensitization profiles, is associated with the diagnosis and symptom elicitation of asthma and allergic rhinitis, and in some studies with decreased lung function and with a diagnosis of atopic dermatitis.
Cross reactivity
Fel d 7 exhibits clinically relevant cross-reactivity with related lipocalin Can f 1 from dog. In cat-sensitized patients, Fel d 7 cross-reactivity prevents the use of Can f 1 as a marker allergen for genuine dog sensitization.
Author: Dr. Joana Vitte
Reviewed by: Dr. Magnus Borres
References
1. van Hage M, Kack U, Asarnoj A, Konradsen JR. An update on the prevalence and diagnosis of cat and dog allergy - Emphasizing the role of molecular allergy diagnostics. Mol Immunol. 2023;157:1-7.
2. Schoos AM, Nwaru BI, Borres MP. Component-resolved diagnostics in pet allergy: Current perspectives and future directions. J Allergy Clin Immunol. 2021;147(4):1164-73.
3. Roger A, Lazo C, Arias N, Quirant B, Albert N, Gomez M, et al. Using Component-Resolved Diagnosis to Characterize the Sensitization to Specific Cat and Dog Allergens in Patients with Allergic Respiratory Diseases in Catalonia, Spain. Int Arch Allergy Immunol. 2023;184(5):440-6.
4. Hemmer W, Sestak-Greinecker G, Braunsteiner T, Wantke F, Wohrl S. Molecular sensitization patterns in animal allergy: Relationship with clinical relevance and pet ownership. Allergy. 2021;76(12):3687-96.
5. Dramburg S, Hilger C, Santos AF, de Las Vecillas L, Aalberse RC, Acevedo N, et al. EAACI Molecular Allergology User's Guide 2.0. Pediatr Allergy Immunol. 2023;34 Suppl 28:e13854.
6. Trifonova D, Curin M, Riabova K, Karsonova A, Keller W, Gronlund H, et al. Allergenic Activity of Individual Cat Allergen Molecules. Int J Mol Sci. 2023;24(23).
7. Smith W, O'Neil SE, Hales BJ, Chai TL, Hazell LA, Tanyaratsrisakul S, et al. Two newly identified cat allergens: the von Ebner gland protein Fel d 7 and the latherin-like protein Fel d 8. Int Arch Allergy Immunol. 2011;156(2):159-70.
8. Riabova K, Karsonova AV, van Hage M, Kack U, Konradsen JR, Gronlund H, et al. Molecular Allergen-Specific IgE Recognition Profiles and Cumulative Specific IgE Levels Associated with Phenotypes of Cat Allergy. Int J Mol Sci. 2022;23(13).
9. Ozuygur Ermis SS, Norouzi A, Borres MP, Basna R, Ekerljung L, Malmhall C, et al. Sensitization patterns to cat molecular allergens in subjects with allergic sensitization to cat dander. Clin Transl Allergy. 2023;13(8):e12294.
10. Kang SY, Yang MS, Borres MP, Andersson M, Lee SM, Lee SP. The association between specific IgE antibodies to component allergens and allergic symptoms on dog and cat exposure among Korean pet exhibition participants. World Allergy Organ J. 2022;15(11):100709.
11. Tsolakis N, Malinovschi A, Nordvall L, Mattsson L, Lidholm J, Pedroletti C, et al. Sensitization to minor cat allergen components is associated with type-2 biomarkers in young asthmatics. Clin Exp Allergy. 2018;48(9):1186-94.
12. Eidukaite A, Gorbikova E, Miskinyte M, Adomaite I, Rudzeviciene O, Siaurys A, et al. Molecular sensitization patterns to cat and dog allergens in Lithuanian children population. World Allergy Organ J. 2023;16(10):100827.
13. Veloso JF, Brandao Guedes PE, Lacerda LC, Santana JO, Mora-Ocampo IY, Pirovani CP, et al. Tear Film Proteome of Healthy Domestic Cats. Vet Med Int. 2021;2021:8708023.
14. Apostolovic D, Sanchez-Vidaurre S, Waden K, Curin M, Grundstrom J, Gafvelin G, et al. The cat lipocalin Fel d 7 and its cross-reactivity with the dog lipocalin Can f 1. Allergy. 2016;71(10):1490-5.
15. Schoos AM, Kattan JD, Gimenez G, Sampson HA. Sensitization phenotypes based on protein groups and associations to allergic diseases in children. J Allergy Clin Immunol. 2016;137(4):1277-80.
16. Uriarte SA, Grönlund H, Wintersand A, Bronge J, Sastre J. Clinical and Immunologic Changes due to Subcutaneous Immunotherapy With Cat and Dog Extracts Using an Ultrarush Up-Dosing Phase: A Real-Life Study. J Investig Allergol Clin Immunol. 2022;32(2):133-40.
17. IUIS/WHO. IUIS/WHO Fel d 7 2024 [Available from: https://allergen.org/viewallergen.php?aid=665.
18. Min J, Foo ACY, Gabel SA, Perera L, DeRose EF, Pomes A, et al. Structural and ligand binding analysis of the pet allergens Can f 1 and Fel d 7. Front Allergy. 2023;4:1133412.
19. Hilger C, Janssen-Weets B, Swiontek K. Hypoallergenic animals: A promise of hope for allergic patients? Allergol Select. 2024;8:64-9.