ThermoFisher Scientific
Search
clear search
Recent searches Clear History
clear search
Search
Recent searches Clear History
Contactez-nous
Allergen Encyclopedia
Table of Contents
Allergène complet
f262 Aubergine, eggplant
f262 Aubergine, eggplant Scientific Information
Type:
Whole Allergen
Nom d’affichage:
Aubergine, eggplant
Family:
Solanaceae
Latin Name:
Solanum melongena
Other Names:
Aubergine, Eggplant, Brinjal, Garden egg, Jew’s apple, Mad apple, Pea apple, Egg apple, Guinea squash
Clinical Relevance
IgE-mediated reactions
Aubergine can induce symptoms of food allergy in sensitised individuals (2-5). But food allergy to ingesting the fruit of this plant is uncommon; reactions have been attributed to cross-reactivity with Tomato and grass pollen allergens in 1 individual (6), and with Parietaria pollen allergen in 5 cases (4).
Eggplant allergy following ingestion of the fruit has been described in 5 individuals, who were also sensitised to Wall pellitory (Parietaria) pollen: 3 women and 2 men aged between 24 and 50 years. In 3 cases, symptoms of OAS were reported (only oropharyngeal symptoms in 1; another also had symptoms of cough and dysphonia, and 1 had vomiting). Two had systemic symptoms as well (1 urticaria, 1 anaphylaxis) (4).
In a random survey of 500 individuals in India, 66 (11%) were reported as having Eggplant allergy, based on case history and skin tests. The authors suggest that this high incidence is probably due to the presence of histamine and serotonin in the plant. In this report, 3 cases of allergy following ingestion of Aubergine were described:A 23-year-old woman with itchiness and an unpleasant feeling while eating food prepared with Eggplant. There was immediate rash, and facial oedema in about 20 minutes.A 25-year-old man who developed itching within 15 minutes after eating any food prepared using Eggplant. Within an hour, the rash became prominent, with itching of the throat and hoarseness. An 18-year-old-girl developed itching in the throat and of the skin within 30 minutes of the ingestion of Eggplant. Generalised uricaria with severe pruritis developed within 1 to 2 hours. SPT and prick-to-prick tests were positive in all 3. IgE antibodies to raw and cooked Eggplant were detected (2).
An Indian study reports on allergy caused by ingestion of Eggplant in a 31-year-old man. His allergy to Aubergine had clearly manifested at the age of 10 years, although symptoms had been first noticed by his mother at the age of 6. He experienced itching or discomfort while eating curry or other foods containing Aubergine. Within 1 to 2 hours of consuming foods containing Aubergine, itchy skin rashes appeared all over the body, along with itching of the throat and hoarseness. Skin prick tests were positive with 4 varieties of Eggplant; however, serum Aubergine-specific IgE antibodies were not detected. It was suggested that the causative allergen was a low-molecular-weight non-protein secondary metabolite of less than 1 kDa (7).
A 31-year-old Spanish woman developed anaphylaxis after eating Banana. She had had a previous diagnosis of Latex-fruit syndrome after presenting with rhinitis and skin and pharyngeal pruritus after eating Chestnut. She also experienced angioedema of the face and hands, conjunctivitis, generalised pruritus, glottic oedema and difficulty in breathing immediately after ingesting Aubergine (5).
Eggplant fruit has induced IgE-mediated immediate-type hypersensitivity reactions, such as rhinorrhoea, urticaria, food allergy and asthma (6).
An Indian study evaluated the possible effect of a specific elimination diet on symptoms of 24 children aged 3 to 15 years with documented deterioration in control of their perennial asthma. IgE antibody analysis for a range of food items found that 22 (92%) had IgE antibodies directed at Aubergine (8).
Anaphylaxis to Aubergine in a Latex-allergic 27-year-old female doctor was reported. She experienced generalised itching, dyspnoea, dizziness, vomiting, nausea, abdominal pain, diarrhoea, and rash over the whole body immediately after eating boiled Eggplant. Skin reactivity to raw and cooked Aubergine was detected. IgE antibodies to raw and cooked Aubergine were found, and 1 protein demonstrated a degree of cross-reactivity with a Latex protein (3).
Allergy to Aubergine pollen has been described. Immediate IgE-mediated hypersensitivity reactions (rhinorrhoea and asthma) were described in a 43-year-old man. He presented with rhinoconjunctivitis and a dry cough when working in a greenhouse where Eggplants were cultivated. SPT to Eggplant fruit was negative, but SPT and conjunctival challenge with extract from the flower petals and pollen was positive (9). Similarly, occupational allergy to the plant pollen in 2 commercial gardeners was reported (10)
Other reactions
Contact dermatitis due to Eggplant has been reported (11).
Contact dermatitis was reported in a 28-year-old woman. She had a 3-month history of bilateral hand eczema following home cultivation of Eggplants, Roses and Tomatoes. A patch test to chopped Eggplant leaves was positive. A delayed-type hypersensitivity reaction was suggested (11).
Aubergine is high in histamine, which may result in histamine reactions in susceptible individuals (12).
Aubergine contains the alkaloid alpha-solanine, which is a human plasma cholinesterase inhibitor (13).
Molecular Aspects
No allergens from this plant have yet been characterised, although several IgE-binding Eggplant proteins have been reported (6). Recently, protein bands of 71, 64 and 60 kDa were detected in sera of 3 Indian Eggplant-allergic individuals. The 71 kDa protein appears to be heat-stable (2). The same authors subsequently reported on a 31-year old-individual with allergy caused by ingestion of Eggplant, in whom skin prick testing was positive with 4 varieties of Eggplant but with negative Aubergine IgE antibody levels. Laboratory analysis revealed that the causative allergen was a low-molecular-weight non-protein secondary metabolite of less than 1 kDa (7). The authors point out that possible non-protein but reaction-causing compounds in Eggplant include pigments (cyanidin, delphinidin, lycoxanthin, and nasunin), alkaloids (solamargine, solanidine, solanine, solasodine, solasonine, and trigonelline), and phytosterols (14). The alkaloid properties of Aubergine have been suggested as the cause of occasional sensitivities to Aubergine (6), though direct evidence is still lacking (7).
In a Korean individual who experienced anaphylaxis to Aubergine with cross-reactivity to Latex, 3 IgE-binding proteins were identified from fresh and cooked Aubergine: 1 band between 22 and 36 kDa, 1 band near 36 kDa, and 1 band between 36 and 50 kDa (3).
A lipid transfer protein has been detected in Aubergine (15). In an allergenicity assessment of the related Ethiopian eggplant (Solanum aethiopicum), profilin and lipid transfer proteins have been detected (16).
Cross Reactivity
Eggplant is a member of the Nightshade family; it is closely related to the Potato and the Tomato. An extensive cross-reactivity among the different individual species of the genus could be expected, as well as to a certain degree among members of the family Solanaceae (6,17). This is supported by research reporting that antigenically cross-reactive material from Tobacco leaf could be found in Eggplants, Green peppers, Potatoes, and Tomatoes, which are all members of the family Solanaceae (18).
Allergic reactions to Eggplant in subjects with Parietaria pollen sensitisation have been reported (4).
A lipid transfer protein has been detected in Aubergine, which may result in cross-reactivity with other lipid transfer protein-containing foods (15).
Anaphylaxis to Eggplant was described in a patient with Latex allergy. Further investigation demonstrated that a protein in boiled Eggplant (and to a lesser degree, raw Eggplant) significantly inhibited Latex antigen. One IgE-binding component with the same molecular weight (between 22 and 36 kDa), from Eggplant and Latex, was detected as a candidate for the cross-reactivity; the protein did not display cross-reactivity with Potato (3). The clinical significance of this cross-sensitisation was illustrated in a 31-year-old Spanish woman with Latex allergy who was also allergic to Banana and Eggplant, as shown by case history and a skin prick test (5).
Compiled By
Last reviewed: June 2022
References
  1. Lawande K F, Chavan J K. Eggplant (Brinjal). In: Salunkhe DK, Kadam SS, editors. Handbook of Vegetable Science and Technology: Production, Composition, Storage, and Processing. New York, Marcel Dekker 1998;225-44
  2. Pramod SN, Venkatesh YP. Allergy to eggplant (Solanum melongena) caused by a putative secondary metabolite. J Investig Allergol Clin Immunol 2008;18(1):59-62
  3. Lee J, Cho YS, Park SY, Lee C-K, Yoo B, Moon H-B, Park H-S. Eggplant anaphylaxis in a patient with latex allergy.
    J Allergy Clin Immunol 2004;113(5):995-6
  4. Pucci S, Incorvaia C. Allergic reactions to eggplant in subjects with parietaria [Poster] 8th International Symposium on Problems of Food Allergy, Venice. 2001, March 11-13
  5. Gamboa PM, Sanchez-Monge R, Diaz-Perales A, Salcedo G, Ansotegui J, Sanz ML. Latex-vegetable syndrome due to custard apple and aubergine: new variations of the hevein symphony. J Investig Allergol Clin Immunol 2005;15(4):308-11
  6. Muthiah R, Zondlo A, Zondlo J, Kagen S. Eggplant anaphylaxis: isolation of the major allergens and demonstration of cross-reactivity with other Solanaceae. [Abstract] J Allergy Clin Immunol 1996:97:334
  7. Pramod SN, Venkatesh YP. Allergy to eggplant (Solanum melongena).J Allergy Clin Immunol 2004;113(1):171-3
  8. Agarkhedkar SR, Bapat HB, Bapat BN. Avoidance of food allergens in childhood asthma. Indian Pediatr. 2005; 42(4):362-6
  9. Gil M, Hogendijk S, Hauser C. Allergy to eggplant flower pollen. Allergy 2002;57(7):652
  10. Gerth van Wijk R, Toorenenbergen AW, Dieges PH. Occupational pollinosis in commercial gardeners. [Dutch] Ned Tijdschr Geneeskd 1989;133(42):2081-3
  11. Kabashima K, Miyachi Y. Contact dermatitis due to eggplant.Contact Dermatitis 2004;50(2):101-2
  12. Feldman JM. Histaminuria from histamine-rich foods. Arch Intern Med. 1983;143(11):2099-102
  13. Beier RC. Natural pesticides and bioactive components in foods. Rev Environ Contam Toxicol 1990;113:47-137
  14. Dr. Duke’s Phytochemical & Ethnobotanical Databases USDA-ARS-NGRL, Beltsville Agricultural Research Center, Beltsville, Maryland, USA. www.ars-grin.gov/duke/ 2008
  15. Asero R, Mistrello G, Roncarolo D, Amato S. Detection of some safe plant-derived foods for LTP-allergic patients.
    Int Arch Allergy Immunol 2007;144(1):57-63
  16. Gubesch M, Theler B, Dutta M, Baumer B, Mathis A, Holzhauser T, Vieths S, Ballmer-Weber BK. Strategy for allergenicity assessment of ’natural novel foods’: clinical and molecular investigation of exotic vegetables (water spinach, hyacinth bean and Ethiopian eggplant). Allergy 2007 Nov;62(11):1243-50
  17. Yman L. Botanical relations and immunological cross-reactions in pollen allergy. 2nd ed. Pharmacia Diagnostics AB. Uppsala. Sweden. 1982: ISBN 91-970475-09
  18. Becker CG, Dubin T, Wiedemann HP. Hypersensitivity to tobacco antigen.
    Proc Natl Acad Sci U S A 1976;73(5):1712-6
  19. Patnaik N. Garden of Life: An introduction to the healing plants of India. New Delhi: Aquarian, an imprint of HarperCollins Publishers 1993:106-7