Recent searches Clear History
聯絡我們
Allergen Encyclopedia
Table of Contents
Component
f351 Pen a 1, Tropomyosin, Shrimp
f351 Pen a 1, Tropomyosin, Shrimp Scientific Information
Type:
Component
Name; WHO/IUIS:
Pen a 1, Tropomyosin, Shrimp
Allergen code:
f351
Source Material:
rPen a 1 is a CCD-free recombinant protein
Other Names :
Tropomyosin
Clinical Relevance
Shrimp are small invertebrate marine crustaceans with 10 jointed legs (decapod) on the thorax, well-developed swimmerets on the abdominal segments, and a body that is compressed laterally. They live on the floor of oceans and lakes. There are over 2,000 different species of Shrimp worldwide.
One common commercial Shrimp is of the genus Peneus.
There are several other crustacean forms that are commonly called Shrimp although they do not belong to the same order as the true Shrimp, order Decapoda (phylum Arthropoda, subphylum Crustacea, class Malacostraca), which also includes the Lobsters and Crabs.
Shrimp may be divided into 3 basic categories: cold-water or northern; warm-water, tropical, or southern; and freshwater. However the nomenclature is complicated and the term “Shrimp” sometimes applies to all crustaceans of the Natantia group, regardless of size. The terms “Prawn” and “Scampi” are often used interchangeably with Shrimp.
rPen a 1 from Brown shrimp (P. aztecus), is representative of other shrimp tropomyosin.
Recombinant allergens, which are genetically engineered isoforms resembling allergen molecules from known allergen extracts, have immunoglobulin E (IgE) antibody binding comparable to that of natural allergens and generally show excellent reactivity in in vitro and in vivo diagnostic tests (1). To date, many different recombinant allergens of pollens, molds, mites, bee venoms, foods and latex have been cloned, sequenced, and expressed. Recombinant allergens have a wide variety of uses, from the diagnosis and management of allergic patients to the development of immunotherapy to the standardisation of allergenic test products as tools in molecular allergology.
Recombinant Pen a 1 and natural Pen a 1 are structurally and immunologically identical (2).
At least 13 allergens are found in extracts of cooked Brown shrimp, and the 36-kDa muscle protein tropomyosin (Pen a 1) has been identified as a major Shrimp allergen. It is detected by sera of more than 80% of all subjects allergic to Shrimp and binds up to 75% of all Shrimp-specific IgE antibodies (2-3). Amino acid sequence identities with natural allergenic and non-allergenic tropomyosins ranged from 80% to 99% and 51% to 58%, respectively (2). Since Beef, Pork and Chicken are other tropomyosin-containing foods that are not very allergenic, tropomyosins can serve in investigations of the contribution of the varying structural properties of a protein to its allergenicity (4).
Recombinant allergens are particularly useful in addressing allergies that manifest wide cross-reactivity, such as allergies to crustaceans, Cockroaches and House dust mites (5).
Allergens from Penaeus aztecus listed by IUIS*
Pen a 1
*International Union of Immunological Societies (www.allergen.org) Jan. 2008.
Molecular Aspects
Allergen Description
Recombinant non-glycosylated protein produced in an E. coli strain carrying a cloned cDNA encoding Penaeus aztecus allergen Pen a 1.
Pen a 1 (4,6-7), the major protein of Brown shrimp (Penaeus aztecus), is a muscle protein tropomyosin (5,7-10). The allergen is heat-stable and is found in both raw and cooked Shrimp (11).
Tropomyosin comprises a class of highly conserved proteins with multiple isoforms found in both muscle and nonmuscle cells of all species of vertebrates and invertebrates. It is an abundant and heat-stable protein that constitutes up to 20% of total protein in the edible part of the animal. It is physically associated with actin and myosin in muscIe fibres and other motile filaments. Allergenic tropomyosins are found in invertebrates such as crustaceans (Shrimp, Lobster, Crab, Crawfish), arachnids (House dust mites), insects (Cockroaches), and mollusks (e.g., Squid); but there is a distinct lack of allergenic cross-reactivity between these tropomyosins and those from vertebrates: their tropomyosins  are virtually nonallergenic (11). Tropomyosins present in vertebrate sources of food, e.g., bony fish, Beef, Pork and Chicken, are rarely allergenic to human beings, as compared to Lobster, Shrimp and Cockroach. Moreover, the invertebrate tropomyosins have high IgE cross-reactivity, and have therefore been referred to as panallergens. Tropomyosin is major allergen in crustaceans.  Pen a 1 is quite important: approximately 80% of individuals sensitised to the Brown shrimp were reported to show IgE antibody reactivity to Pen a 1 (6,12).
Studies suggest that IgE-binding epitopes are restricted to certain parts of the Pen a 1 molecule and that Pen a 1 may have several similar epitopes. Pen a 1 epitopes do not appear to be located in the highly homologous parts of the molecule (9).
Studies have demonstrated that tropomyosin is an important allergen in crustaceans other than Shrimp, such as Lobster - both the Spiny lobster Panulirus stimpsoni and the American lobster Homarus americanus (Pan s 1, Hom a 1) (13-14) – the Crab Charybdis feriatus (Cha f 1) (15), mollusks such as the Squid Todarodes pacificus (Tod p 1) (16),  the Snail Turbo cornutus (Tur c 1) (17),  the Oyster Crassostrea gigas (Cra g 1) (18),  and other invertebrates such as the House dust mites Dermatophagoides farinae (Der f 10) and Dermatophagoides pteronyssinus (Der p 10) (19-20), and the Cockroach Periplaneta americana (Per a 7) (21-22). Immunological relationships based on tropomyosin have also been demonstrated between crustaceans, Cockroaches and House dust mites, suggesting that tropomyosin is an important cross-sensitising panallergen (5).
In further evidence, rPen a 1 has been shown to extensively and specifically compete for IgE binding to extracts of other crustacean species, House dust mites and the German cockroach (6). The tropomyosins Pan s 1 (Spiny lobster, Panulirus stimpsoni) and Hom a 1 (American lobster, Homarus americanus) have been shown to have significant homology to Shrimp tropomyosin (14).
Nonetheless, studies suggest that there may be species-specific allergens in shrimps. In a comparision of the allergens from the edible Shrimp species Penaeus setifecus (White shrimp) and Penaeus aztecus (Brown shrimp) in 31 individuals with a history of immediate hypersensitivity reactions after Shrimp ingestion, skin-specific IgE to both types of extract was observed in 77% (23/30) of the subjects; 1 individual reacted to Brown shrimp extract only. Serum-specific IgE to both extracts was demonstrated in 16/31 study participants; 1 subject reacted only to White shrimp extract, and 2 subjects to Brown shrimp extract alone. Species specificity is important because it may explain the intermittent symptoms of some study subjects (23). 
Recombinant Pen a 1 has been demonstrated to have allergenic activity not only similar to that of its own native allergen (6), but also very similar to that of the Greasyback shrimp (Metapenaeus ensis) tropomyosin (Met e 1) (7). Four recombinant, IgE-reactive Pen a 1 peptides, isolated in a study, showed various degrees of sequence identity with tropomyosins of other arthropods such as fruitfly (Drosophila melanogaster), House dust mite, helminths and vertebrates (4,7,11).
Tropomyosin from the Mite Blomia tropicalis (Blo t 10) was demonstrated to have cross-reactivity with Der p 10 of Dermatophagoides pteronyssinus, and to share up to 96% amino acid identity to tropomyosin of other Mites. Although Blo t 10 and Der p 10 were shown to be significantly cross-reactive, unique IgE epitopes do exist (24).
Tropomyosin has also been described in arthropods, namely Fly (Musca domestica), Moth (Ephestia spp.) and Spider (Tegenaria spp.). In a study of 100 patients allergic to household arthropods, cross-reactivity due to tropomyosin was demonstrated in a large variety of extracts obtained from insects, mites, crustaceans, mollusks and parasites (25).
While tropomyosin is a major factor in food allergy to invertebrates, it plays a less prominent role in inhalation allergy to Mites and Cockroaches. Tropomyosin has been isolated from Anisakis but does not appear to be an important allergen in Anisakis sensitisation (26). Similarly, Tur c 1, the tropomyosin from the gastropod Turbo cornutus, has an IgE-binding epitope that is dissimilar to those proposed for Cra g 1 from the Oyster Crassostrea gigas, and to Pen i 1 from the Shrimp Penaeus indicus (17).
Clinical and serological reactivity to both Mites and Snails has been described, and the development of sensitisation and allergic symptoms to Snail and Shrimp following immunotherapy treatment with Mite extract has been reported (6). IgE antibody reactivity to Shrimp can occur in an unexposed population of individuals; a study of Orthodox Jews unexposed to Shrimp demonstrated that some subjects allergic to HDM and/or Cockroach showed substantial IgE antibody reactivity to the Shrimp tropomyosin Pen a 1. Based on inhibition with Cockroach and/or Dust mite extracts, this reactivity appeared to be due to cross-reacting tropomyosins (27).
Therefore, as IgE-mediated food allergy to crustaceans and mollusks is relatively common, and affected individuals typically react to a range of different species, tropomyosin sensitivity may be useful as a diagnostic marker for allergic sensitisation to invertebrate foods. rPen a 1 has potential use as a diagnostic reagent to determine not only sensitisation specifically to Brown shrimp, but also sensitisation to and cross-reactivity with tropomyosin allergens from other species.
Compiled By
Last reviewed: June 2022.
References
  1. Niederberger V, Stubner P, Spitzauer S, Kraft D, Valenta R, Ehrenberger K, Horak F. Skin test results but not serology reflect immediate type respiratory sensitivity: a study performed with recombinant allergen molecules.
    J Invest Dermatol 2001;117(4):848-51
  2. Reese G, Schicktanz S, Lauer I, Randow S, Luttkopf D, Vogel L, Lehrer SB, Vieths S. Structural, immunological and functional properties of natural recombinant Pen a 1, the major allergen of Brown Shrimp, Penaeus aztecus.
    Clin Exp Allergy 2006;36(4):517-24
  3. Jeoung BJ, Reese G, Hauck P, Oliver JB, Daul CB, Lehrer SB. Quantification of the major brown shrimp allergen Pen a 1 (tropomyosin) by a monoclonal antibody-based sandwich ELISA. J Allergy Clin Immunol 1997;100(2):229-34
  4. Reese G, Jeoung BJ, Daul CB, Lehrer SB. Characterization of recombinant shrimp allergen Pen a 1 (tropomyosin). Int Arch Allergy Immunol 1997;113(1-3):240-2
  5. Reese G, Ayuso R, Lehrer SB. Tropomyosin: an invertebrate pan-allergen. Int Arch Allergy Immunol 1999;119(4):247-58
  6. Marknell DeWitt A, Mattsson L, Lauer I, Reese G, Lidholm J. Recombinant tropomyosin from Penaeus aztecus (rPen a 1) for measurement of specific immunoglobulin E antibodies relevant in food allergy to crustaceans and other invertebrates Molec Nutr & Food Res 2004;48(5):370-9
  7. Daul CB, Slattery M, Morgan JE, Lehrer SB. Common crustacea allergens: identification of B cell epitopes with the shrimp specific monoclonal antibodies. In: “Molecular Biology and Immunology of Allergens” (D. Kraft and A. Sehon, eds.). CRC Press, Boca Raton 1993;291-3
  8. Reese G, Daul CB, Lehrer SB. Antigenic analysis (IgE and monoclonal antibodies) of the major shrimp allergen Pen a 1 (Tropomyosin) from Penaeus aztecus. Int Arch Allergy Immunol 1995;107(1-3):245-7
  9. Reese G, Tracey D, Daul CB, Lehrer SB. IgE and monoclonal antibody reactivities to the major shrimp allergen Pen a 1 (tropomyosin) and vertebrate tropomyosins.
    Adv Exp Med Biol 1996;409:225-30
  10. Daul CB, Slattery M et al. Identification of the major brown shrimp (Penaeus aztecus) allergen as the muscle protein tropomyosin. Int Arch Allergy Immunol 1994;105(1):49-55
  11. Shanti KN, Martin BM, Nagpal S, Metcalfe DD, Rao PV. Identification of tropomyosin as the major shrimp allergen and characterization of its IgE-binding epitopes.
    J Immunol 1993 Nov 15;151(10):5354-63
  12. Ayuso R, Lehrer SB, Reese G. Identification of continuous, allergenic regions of the major shrimp allergen pen a 1 (tropomyosin). Int Arch Allergy Immunol 2002;127(1):27-37
  13. Mykles DL, Cotton JL, Taniguchi H, Sano K, Maeda Y. Cloning of tropomyosins from lobster (Homarus americanus) striated muscles: fast and slow isoforms may be generated from the same transcript.
    J Muscle Res Cell Motil 1998;19(2):105-15
  14. Leung PS, Chen YC, et al. Molecular identification of the lobster muscle protein tropomyosin as a seafood allergen.
    Mol Mar Biol Biotechnol 1998;7(1):12-20
  15. Leung PS, Chen YC, Gershwin ME, Wong SH, Kwan HS, Chu KH. Identification and molecular characterization of Charybdis feriatus tropomyosin, the major crab allergen. J Allergy Clin Immunol 1998;102(5):847-52
  16. Miyazawa H, Fukamachi H, Inagaki Y, Reese G, Daul CB et al. Identification of the first major allergen of a squid (Todarodes pacificus).
    J Allergy Clin Immunol 1996;98(5 Pt 1):948-53
  17. Ishikawa M, Ishida M, Shimakura K, Nagashima Y, Shiomi K. Purification and IgE-binding epitopes of a major allergen in the gastropod Turbo cornutus. Biosci Biotechnol Biochem 1998;62(7):1337-43
  18. Leung PS, Chu KH. cDNA cloning and molecular identification of the major oyster allergen from the Pacific oyster Crassostrea gigas.
    Clin Exp Allergy 2001;31(8):1287-94
  19. Aki T, Kodama T, Fujikawa A, Miura K, Shigeta S, Wada T, Jyo T, Murooka Y, Oka S, Ono K. Immunochemical characterization of recombinant and native tropomyosins as a new allergen from the house dust mite, Dermatophagoides farinae.
    J Allergy Clin Immunol 1995;96(1):74-83
  20. Asturias JA, Arilla MC, Gomez-Bayon N, Martinez A, Martinez J, Palacios R. Sequencing and high level expression in Escherichia coli of the tropomyosin allergen (Der p 10) from Dermatophagoides pteronyssinus. Biochim Biophys Acta 1998;1397(1):27-30
  21. Asturias JA, Gomez-Bayon N, Arilla MC, Martinez A, Palacios R, Sanchez-Gascon F, Martinez J. Molecular characterization of American cockroach tropomyosin (Periplaneta americana allergen 7), a cross-reactive allergen.
    J Immunol 1999;162(7):4342-8
  22. Santos ABR, obias KR, Ferriani VPL et al. Identification of tropomyosin from Periplaneta americana as a major cockroach allergen. J Allergy Clin Immunol 1999;103:S122
  23. Morgan JE, O’Neil CE, Daul CB, Lehrer SB. Species-specific shrimp allergens: RAST and RAST-inhibition studies. J Allergy Clin Immunol 1989;83(6):1112-7
  24. Yi FC, Cheong N, Shek PC, Wang DY, Chua KY, Lee BW. Identification of shared and unique immunoglobulin E epitopes of the highly conserved tropomyosins in Blomia tropicalis and Dermatophagoides pteronyssinus.
    Clin Exp Allergy 2002;32(8):1203-10
  25. Martinez A, Martinez J, Palacios R, Panzani R. Importance of tropomyosin in the allergy to household arthropods. Cross-reactivity with other invertebrate extracts. Allergol Immunopathol (Madr) 1997;25(3):118-26
  26. Asturias JA, Eraso E, Moneo I, Martinez A.
    Is tropomyosin an allergen in Anisakis?
    Allergy 2000;55(9):898-9
  27. Fernandes J, Reshef A, Patton L, Ayuso R, Reese G, Lehrer S. Immunoglobulin E antibody reactivity to the major shrimp allergen, tropomyosin, in unexposed Orthodox Jews. Clin Exp Allergy 2003;33(7):956-61