EVOS cell imaging systems are designed to eliminate the complexities of capturing multicolor fluorescent images without compromising on performance. These units combine all features of a digital inverted microscope workstation into a single, compact device that can be mastered in minutes. Even the most novice users will be able to spend more time analyzing data rather than worrying about microscope operation. Whether you’re capturing exquisite images for publication or counting cells for viability determination, these systems are designed to be easier, smarter, and faster.
Which system is right for you?
Thermo Fisher Scientific offers a fluorescence imaging solution to fit your application—you choose the system that is right for you based on the imaging applications you want to run. Any or all of the following could influence your choice of platform:
  • Imaging in the cell culture hood
  • Number of excitation channels
  • Onstage incubator with environmental control
  • Automated cell counting
EVOS imaging systems brochure
See the entire portfolio of easy to use cell imaging instruments that can be configured with a variety of objectives, light cubes, and vessel holders to meet your research needs.
Download the EVOS brochure
Use the selection guide below to find a system that fits you.
Fluorescence imaging selection guide
  EVOS FLoid
EVOS FLoid
Perfect for quick fluorescence visualization Virtual tour
EVOS FL
EVOS FL
Perfect for multichannel fluorescence imaging and transfections
Virtual tour
EVOS FL Auto
EVOS FL Auto
Perfect for a variety of advanced automated applications
Virtual tour
High-resolution LCD display 15" 15" 22"
Available objectives 20x fixed 1.25x–100x 1.25x–100x
Camera Monochrome CCD Choice of color or monochrome CCD Dual monochrome (CCD) and color (CMOS)
Transmitted light (bright-field & phase-contrast)
Epifluorescence imaging
Fluorescent channels 3
(DAPI, GFP, Texas Red)
4
(changeable light cubes)
4
(changeable light cubes)
Fits in tissue culture hood    
Time-lapse imaging  
Onstage incubator    
(optional)
Autofocus    
High-resolution image mapping    
Cell counting  
Weight 11.8 kg (26.0 lb) 15.3 kg (33.7 lb) 20 kg (44.1 lb)
Dimensions    H: 53.6 cm (21.1")
   W: 40.4 cm (15.9")
   D: 35.3 cm (13.9")
   H: 57.8 cm (22.8")
   W: 35.5 cm (14.0")
   D: 47.0 cm (18.5")
   H: 32.2 cm (12.7")
   W: 34.3 cm (13.5")
   D: 47.2 cm (18.6")