Case Study
COVID-19 Positive Rule-out Bacterial Pneumonia
Critical Care Medicine Physician
New York, USA
Learn more about implementing procalcitonin testing
in your hospital.
Explore similar topics
Key Takeaways
Takeaway #1
Takeaway #1
Bacterial co-infection and secondary bacterial infections are relatively infrequent (< 10%) in hospitalized patients with COVID-19, yet over 70% of patients receive antibiotics on hospital admission and often for several days.1
Takeaway #2
Takeaway #2
Majority of these patients may not require empiric IV antibiotics. Procalcitonin has a high negative-predictive value at ≤ 0.25 μg/L for determining need for antibiotics in patients with community-acquired pneumonia, including those who have underlying malignancy.
Takeaway #3
Takeaway #3
PCT levels also have a high negative predictive value when compared to blood cultures.2,3 Serial PCTs can assist with confirming the initial negative result and indicate that antibiotics are no longer necessary.
Share your Clinical Experience
Submit your own PCT Case Study and you may be selected to be featured on procalcitonin.com. 
Submit a case study here
Share your experience
Related Resources
References
  1. Langford BJ, So M, Raybardhan S, Leung V, Westwood D, MacFadden DR, Soucy JR, Daneman N. Bacterial co-infection and secondary infection in patients with COVID-19: a living rapid review and meta-analysis. Clin Microbiol Infect. 2020 Dec;26(12):1622-1629. doi: 10.1016/j.cmi.2020.07.016. Epub 2020 Jul 22. PMID: 32711058.
  2. Riedel S, Melendez JH, An AT, Rosenbaum JE, Zenilman JM. Procalcitonin as a marker for the detection of bacteremia and sepsis in the emergency department. Am J Clin Pathol. 2011 Feb;135(2):182-9.
  3. Oussalah A, Ferrand J, Filhine-Tresarrieu P, Aissa N, Aimone-Gastin I, Namour F, et al. Diagnostic accuracy of procalcitonin for predicting blood culture results in patients with suspected bloodstream infection: an observational study of 35,343 consecutive patients (A STROBE-Compliant Article). Medicine (Baltimore). 2015 Nov;94(44):e1774. 
Back to Top Arrow