Our protein gels welcome packs contain the components you need for outstanding protein separation- from the gel running tank to the protein ladder, we have you covered.
Most protein gel welcome packs provide all of the necessary gels, buffers, and reagents you need as well as our Mini Gel Tank.
Ordering informationFind the right gel for your protein
Gel chemistryBolt Bis-Tris PlusNuPAGE Bis-TrisNovex Tris-GlycineNuPAGE Tris-AcetateNovex Tricine
Molecular Weight rangeBroad MW (6–250 kDa)Broad MW (6–250 kDa)Broad MW (6–250 kDa)High MW (40–500 kDa)Low MW (2.5–40 kDa)
Gel formatMiniMidi
Mini
Midi
Mini
Midi
Mini
Mini
WedgeWell format MiniMidiMidi and miniMidi and mini
Reduced protein degradationXX X 
Fast run timesXX   
Extended shelf lifeXXX  
High loading capacity meets high sample throughput
WedgeWell format Midi protein gels feature wedge-shaped wells that double the sample loading capacity. WedgeWell format Midi gel welcome packs include a SureLock Tandem Tank and offer:
Wedgewell icon
WedgeWell format wells
—load up to 100 µL of samples
Protein gel icon
Wide format gels
—run up to 26 samples per gel
pipette icon
Easy loading
—no specialized pipette tips needed
Ordering Information
Mini gel welcome pack options
Four gel chemistries for tailored protein separations.
Bis-Tris and Tris-glycine are excellent for broad molecular weight range separation, and both feature a long shelf-life. The neutral pH of the Bis-Tris and the Bis-Tris Plus gel chemistries helps preserve protein integrity and offer fast run times.
The Invitrogen Novex Tris-Glycine Gels offer maximum protein separation and crisp, straight bands. NuPAGE Tris-Acetate Gels are recommended for the separation of high molecular weight proteins up to 500 kDa. Novex Tricine Gels are designed for the separation of low molecular weight proteins and peptides.
Wedgewell icon
WedgeWell format wells
—load up to 100 µL of samples
Rocket icon
Run your gel in just 20 minutes—
Bolt Bis-Tris Plus gels
Target icon
Gel chemistry tailored to your target protein
Gel tank, transfer modules, box of precast gels, bottles of buffers and reagents for protein electrophoresis.
Find an excellent protein gel chemistry for your research needs
Decision tree for choosing a protein gel chemistry welcome pack for the right application and research needs
 Click image to enlarge
The choice of whether to use one chemistry or another depends on the abundance of the target protein, the size of the protein, and the downstream application.
Protein Gel Performance Guarantee
Invitrogen precast protein gels are designed and tested to deliver excellent performance, reliability, reproducibility, and consistency of quality. We guarantee it. Download our Certificate of Analysis for more details. No other supplier conducts such stringent quality testing.
Hero banner dark gradient overlay
Stylesheet for Classic Wide Template adjustments
Stylesheet for Classic Wide Template adjustments
For Research Use Only. Not for use in diagnostic procedures.