Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
PEP-235 is a 9 amino acid synthetic peptide corresponding to amino acid residues 436-441 from human Versican. The sequence of this peptide is (amino to carboxy terminus): C G G D (436) P E A A E (441).
This peptide may be used for neutralization and control experiments with the polyclonal antibody that reacts with this product, catalog # PA1-1748A. Using a solution of peptide of equal volume and concentration to the corresponding antibody will yield a large molar excess of peptide (~70-fold) for competitive inhibition of antibody-protein binding reactions.
Reconstitute with 0.1 mL of distilled water.
Versican is a member of the family of large aggregating proteoglycans (also including aggrecan, brevican, and neurocan). The protein shows wide tissue distribution which includes fibrous, articular, and elastic cartilages, as well as nervous, epidermal, arterial, and loose connective tissues. Versican V1 has been shown to enhance cell proliferation, and also induce cell transformation and protect cells from apoptosis. This was demonstrated through its ability to downregulate the expression of proapoptotic Bad. Versican's existence as a extracellular matrix protein in human airways has also lead to the discovery of an inflammatory role in asthmatics.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Chondroitin sulfate proteoglycan 2; Chondroitin sulfate proteoglycan core protein 2; GHAP; Glial hyaluronate-binding protein; Large fibroblast proteoglycan; PG-M; PGM; V1 Neo; Versican core protein; versican proteoglycan; Versican V0; WGN 1
Gene Aliases: CSPG2; ERVR; GHAP; PG-M; VCAN; WGN; WGN1
UniProt ID: (Human) P13611
Entrez Gene ID: (Human) 1462
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support