cleaved D4 GDI Antibody (MA1-41368) in FACS
Intracellular flow cytometric analysis of cleaved D4-GDI in untreated (left) and staurosporine-treated (right) Ramos cells using 0.5 ug of cleaved D4 GDI monoclonal antibody (Product ## MA1-41368). Green represents isotype control; red represents cleaved D4 GDI monoclonal antibody (Product ## MA1-41368).
Intracellular flow cytometric analysis of cleaved D4-GDI in untreated (left) and staurosporine-treated (right) Ramos cells using 0.5 ug of cleaved D4 GDI monoclonal antibody (Product ## MA1-41368). Green represents isotype control; red represents cleaved D4 GDI monoclonal antibody (Product ## MA1-41368).
Related Products
Related Images