Thermo Scientific offers a variety of Class II, Type A2 Biosafety Cabinets that meet your requirements for your application throughout the world. Resource materials are available for your support. Browse our complete portfolio or connect with an expert to help you make a selection.
Class II Biosafety Cabinets [EN 12469]
- Meet the requirements of EN 12469, the European standard for design, construction and performance of Class II biosafety cabinets (BSCs).
- EN 12469 is the generally required standard for BSCs in Europe as well as being frequently required and recommended worldwide.
- Applications for work with hazardous drugs, CMR (carcinogenic, mutagenic, reprotoxic) substances or other specialized procedures may require B2 Biosafety Cabinet or Cytotoxic Safety Cabinets (DIN 12980).
Our featured product, the Herasafe 2030i, innovates optimizing essential environmental protection and contamination prevention, with enhanced user convenience, to deliver the most technologically advanced Class II cabinet,. The 2030i provides unmatched performance accuracy to support demanding cell culture applications such as stem cells and primary cell culturing, and other challenging workflows requiring assured safety.
Compare the features to help with your selection
Select highlighted terms for further definition
Herasafe 2030i |
Safe 2020 |
MSC Advantage |
|
![]() |
![]() |
![]() |
|
| Description | Outstanding protection, comfort and performance for demanding applications with intuitive interface and connectivity. | Outstanding protection, comfort and performance for demanding applications. | Outstanding protection, comfort and performance in an economical design. |
| Protection | → SmartFlow Plus → Cross beam UV Plus → Full text messaging for alarms and alerts |
→ SmartFlow → Cross beam UV |
→ SmartFlow → UV with saved cycle times |
| Ergonomics | → Motorized window with one button start up/shut down → Transparent side walls → Individually selectable tones for alert and alarms |
→ Motorized window → Transparent side walls → SmartClean Plus |
→ Counterbalanced manual window → SmartClean |
| Design | → Over-the-grill armrests → Stainless steel interior (Type 316) |
→ Over-the-grill armrests → Stainless steel interior (type 316) |
→ Over-the-grill armrests → SmartCoat interior |
| Efficiency | → Dual DC intelligent motors → LED lights → Standby mode for further energy savings |
→ Dual DC intelligent motors → Fluorescent lights → Standby mode for further energy savings |
→ Dual DC intelligent motors → Fluorescent lights → Standby mode for further energy savings |
| Download Brochure › |
| Counterbalanced Manual Window |
| Counterbalanced Manual Window |
MSC Advantage
The Thermo Scientific MSC-Advantage meets EN 12469 for a Class II biosafety cabinet. These energy efficient cabinets provide both personal and product protection from biological hazards and contamination and can be connected to external exhaust to provide protection for volatile toxic chemicals used in the cabinet.
- SmartFlow compensation to maintain inflow and the overall balance of inflow and downflow.
- Counter balanced window for easy operation, negatively pressurized double sided walls for greater containment.
- SmartClean providing easy access to the upper portions of cabinet interior through extended window movement.
- Available with 20cm front aperture and SmartCoat interior.
| Size/Internal Width | 0.9m | 1.2m | 1.5m | 1.8m | |
| Exterior dimensions2 (W x H x D) |
mm | 1000 x 1522 x 798 | 1300 x 1522 x 798 | 1600 x 1522 x 798 | 1900 x 1522 x 798 |
| in | 39.4 x 61.8 x 31.5 | 51.2 x 61.8 x 31.5 | 63.0 x 61.8 x 31.5 | 74.8 x 61.8 x 31.5 | |
| Interior dimensions (W x H x D) | mm | 900 x 780 x 630 | 1200 x 780 x 630 | 1500 x 780 x 630 | 1800 x 780 x 630 |
| in | 35.4 x 30.7 x 24.8 | 47.2 x 30.7 x 24.8 | 59.1 x 30.7 x 24.8 | 70.9 x 30.7 x 24.8 | |
| Shipping dimensions1 (W x H x D) | mm | 1110 x 1730 x 925 | 1410 x 1730 x 925 | 1710 x 1730 x 925 | 2010 x 1730 x 925 |
| in | 43.7 x 68.1 x 36.4 | 55.5 x 68.1 x 36.4 | 67.3 x 68.1 x 36.4 | 79.1 x 68.1 x 36.4 | |
| Working height of front window | 200mm (8 inches) | ||||
|
Maximum opening height of front window
|
535mm (21 inches) | ||||
1 Export packaging dimensions and weight upon
2 Exterior height to top of exhaust HEPA filter for clearance purposes is 1500 mm
| Size/Internal Width | 0.9m | 1.2m | 1.5m | 1.8m | |
| Net weight | kg | 170 | 200 | 230 | 280 |
| lbs | 375 | 441 | 507 | 617 | |
| Shipping weight1 | kg | 190 | 225 | 265 | 315 |
| lbs | 419 | 496 | 573 | 694 | |
| Max. load capacity of single segmented work surface per 30 cm segments | kg | 25 | |||
| lbs | 55 | ||||
1 Export packaging dimensions and weight request
| Size/ Internal Width | 0.9m | 1.2m | 1.5m | 1.8m |
| Voltage (V) | 230 | |||
| Frequency (Hz) | 50/60 | |||
| Protection Class | I / IP 20 | |||
| Filter specification | H14 HEPA EN 1822, 99.995% @ MPPS | |||
| Certifications to EN 12469 | TÜV, LNE | TÜV, LNE | TUV | TUV |
| Size/ Internal Width | 0.9m | 1.2m | 1.5m | 1.8m | |
| Lighting power | lux | >800 | >850 | >1250 | >1300 |
| fc | 74 | 79 | 116 | 121 | |
| Sound pressure level (dBA) per EN 12469 | <55 | <55 | <59 | <59 | |
| SmartPort | Two 76 mm (3") plugged cable ports, one on each wall | ||||
| Receptacles4 | 2 sockets on right rear wall | 2 sockets on right and left rear wall | |||
| Service Valves | Up to 4 may be installed through access ports (2 on each side) |
||||
4 The receptacles have a load capacity of up to 5 A and are protected with T 5 A fuses. When all receptacles are in use simultaneously they must not exceed the maximum total load capacity of 5A
| Size/ Internal Width | 0.9m | 1.2m | 1.5m | 1.8m | |
| Energy consumption, operating set point6 (W) | 150 | 200 | 280 | 340 | |
| Energy consumption, Stand-by mode (W) | 40 | 40 | 70 | 70 | |
| Heat output, operating mode (BTU/hr) | 512 | 682 | 955 | 1160 | |
| Heat output, Stand-by mode (lights off, non-vented) (BTU/hr) | 136 | 136 | 239 | 239 | |
| Exhaust/inflow air volume | m3/hr | 292 | 389 | 486 | 583 |
| cfm | 172 | 229 | 286 | 343 | |
| Exhaust volume, thimble ducted | m3/hr | 379 | 505 | 632 | 758 |
| cfm | 223 | 297 | 372 | 446 | |
| Request a quote | 51028225 › | 51025411 › | 51028226 › | 51025413 › | |
6 Clean filters, fans operating at nominal set point, interior lighting activated
Herasafe KS and Safe 2020
The Thermo Scientific Herasafe KS and Safe 2020 meet EN 12469 for a Class II biosafety cabinet. These energy efficient cabinets provide both personal and product protection from biological hazards and contamination and can be connected to external exhaust to provide protection for volatile toxic chemicals used in the cabinet.
- SmartFlow compensation to maintain inflow and the overall balance of inflow and downflow.
- Motorized window for easy operation at cabinet and available handheld controller (Herasafe KS).
- Transparent side walls with Type 316 stainless steel interior.
- Available with 20cm (Safe 2020) or 25cm (Herasafe KS) front aperture.
- SmartClean Plus providing maximum access to the cabinet interior through hinged window movement.
| Size/ Internal Width | 0.9m | 1.2m | 1.5m | 1.8m | |
| Exterior dimensions (W x H x D) | mm | 1000 x 1586 x 799 | 1300 x 1586 x 799 | 1600 x 1586 x 799 | 1900 x 1586 x 799 |
| in | 39.4 x 62.4 x 31.5 | 51.2 x 62.4 x 31.5 | 63.0 x 62.4 x 31.5 | 74.8 x 62.4 x 31.5 | |
| Interior dimensions (W x H x D) | mm | 900 x 780 x 630 | 1200 x 780 x 630 | 1500 x 780 x 630 | 1800 x 780 x 630 |
| in | 35.4 x 30.7 x 24.8 | 47.2 x 30.7 x 24.8 | 59.1 x 30.7 x 24.8 | 70.9 x 30.7 x 24.8 | |
| Shipping dimensions1 (W x H x D) | mm | 1110 x 1760 x 925 | 1410 x 1760 x 925 | 1710 x 1760 x 925 | 2010 x 1760 x 925 |
| in | 43.7 x 69.3 x 36.4 | 55.5 x 69.3 x 36.4 | 67.3 x 69.3 x 36.4 | 79.1 x 69.3 x 36.4 | |
| Herasafe KS - Working height of front window | 250mm (10 inches) |
||||
| Safe 2020 - Working height of front window | 200mm (8 inches) |
||||
| Maximum opening height of front window | 773mm (30.4 inches) |
||||
1 Export packaging dimensions and weight upon request
| Size/ Internal Width | 0.9m | 1.2m | 1.5m | 1.8m | |
| Net weight | kg | 170 | 200 | 230 | 280 |
| lbs | 375 | 441 | 507 | 617 | |
| Shipping weight1 | kg | 190 | 225 | 260 | 315 |
| lbs | 419 | 496 | 573 | 694 | |
| Max. load capacity of one-piece work surface | kg | 50 | |||
| lbs | 110 | ||||
| Max. load capacity of single segmented work surface per 30cm segments3 | kg | 25 | |||
| lbs | 55 | ||||
1 Export packaging dimensions and weight upon request
3 The total load on overall working area must not exceed 50 kg
| Size/ Internal Width | 0.9m | 1.2m | 1.5m | 1.8m |
| Voltage (V) | 230 | |||
| Frequency (Hz) | 50/60 | |||
| Protection Class | I / IP 20 | |||
| Filter specification | H14 HEPA EN 1822, 99.995% @ MPPS | |||
| Herasafe KS Certifications | TÜV | TÜV, LNE | TÜV | TÜV |
| Safe 2020 Certifications | TÜV | TÜV | ||
| Size/ Internal Width | 0.9m | 1.2m | 1.5m | 1.8m | |
| Lighting power | lux | >800 | >850 | >1250 | >1300 |
| fc | >74 | >79 | >116 | >121 | |
| Sound pressure level (dbA) per EN 12469 for Herasafe KS | <58 | <58 | <60 | <60 | |
| Sound pressure level (dbA) per EN 12469 for Safe 2020 | <56 | <56 | <58 | <58 | |
| Receptacles4 | 2 sockets on each side wall | ||||
| Service Valves | Up to 4 may be installed through access ports (2 on each side) Up to 6 more may be factory installed on rear wall (optional) |
||||
4 The receptacles have a load capacity of up to 5 A and are protected with T5A fuses. When all receptacles are in use simultaneously they must not exceed the maximum total load capacity of 5A
| Size/ Internal Width | 0.9m | 1.2m | 1.5m | 1.8m | |
| Energy consumption, operating set point6 (W) | 170 | 210 | 275 | 350 | |
| Energy consumption, Stand-by mode (W) | 40 | 40 | 70 | 70 | |
| Heat output, operating mode (non-vented) (BTU/hr) | 580 | 717 | 938 | 1194 | |
| Heat output, Stand-by mode (lights off, non-vented) (BTU/hr) | 136 | 136 | 239 | 239 | |
| Exhaust/inflow air volume | m3/h | 366 | 490 | 613 | 737 |
| cfm | 215 | 288 | 361 | 434 | |
| Ducting volume requirement, thimble connection | m3/h | 476 | 637 | 797 | 958 |
| cfm | 280 | 375 | 469 | 564 | |
| Request a quote | 51022481 › | 51022482 › | 51022483 › | 51022484 › | |
6 Clean filters, fans operating at nominal set point, interior lighting activated
| Size/ Internal Width | 0.9m | 1.2m | 1.5m | 1.8m | |
| Energy consumption, operating set point6 (W) | 160 | 200 | 265 | 340 | |
| Energy consumption, Stand-by mode (W) | 40 | 40 | 70 | 70 | |
| Heat output, operating mode (non-vented) (BTU/hr) | 546 | 682 | 904 | 1161 | |
| Heat output, Stand-by mode (lights off, non-vented) (BTU/hr) | 136 | 136 | 239 | 239 | |
| Exhaust/inflow air volume (m3/h) | m3/h | 288 | 386 | 483 | 580 |
| cfm | 170 | 227 | 284 | 341 | |
| Ducting volume requirement, thimble connection (m3/h) | m3/h | 375 | 501 | 628 | 754 |
| cfm | 221 | 295 | 370 | 444 | |
| Request a quote | 51026637 › | 51026638 › | 51026639 › | 51026640 › | |
6 Clean filters, fans operating at nominal set point, interior lighting activated
Herasafe 2030i
The Thermo Scientific Herasafe 2030i meets EN 12469 for a Class II biosafety cabinet. These energy efficient cabinets provide both personal and product protection from biological hazards and contamination and can be connected to external exhaust to provide protection for volatile toxic chemicals used in the cabinet.
- SmartFlow Plus compensation to independently maintain and monitor inflow and downflow.
- Graphical User Interface with available one button start up/shut down and enhanced connectivity.
- Motorized window for easy operation, transparent side walls and Type 316 stainless steel interior.
- SmartClean Plus providing maximum access to the cabinet interior through hinged window movement.
| Size/Width | 0.9m | 1.2m | 1.5m | 1.8m | |
| Exterior dimensions (W x H x D) |
mm | 1000 x 1536 x 800 | 1300 x 1536 x 800 | 1600 x 1536 x 800 | 1900 x 1536 x 800 |
| in | 39.4 x 62.4 x 31.5 | 51.2 x 62.4 x 31.5 | 63.0 x 62.4 x 31.5 | 74.8 x 62.4 x 31.5 | |
| Interior dimensions (W x H x D) |
mm | 900 x 780 x 630 | 1200 x 780 x 630 | 1500 x 780 x 630 | 1800 x 780 x 630 |
| in | 35.4 x 30.7 x 24.8 | 47.2 x 30.7 x 24.8 | 59.1 x 30.7 x 24.8 | 70.9 x 30.7 x 24.8 | |
| Shipping dimensions1 (W x H x D) |
mm | 1110 x 1710 x 925 | 1410 x 1710 x 925 | 1710 x 1710 x 925 | 2010 x 1710 x 925 |
| in | 43.7 x 67.3 x 36.4 | 55.5 x 67.3 x 36.4 | 67.3 x 67.3 x 36.4 | 79.1 x 67.3 x 36.4 | |
| Working height of front window | 200 (250mm can be optioned) 8 inches (10 inches can be optioned) |
||||
|
Maximum opening height of front window
|
773mm (30.4 inches) | ||||
1 Export packaging dimensions and weights upon request
| Size/Width | 0.9m | 1.2m | 1.5m | 1.8m | |
| Net weight | kg | 170 | 200 | 230 | 280 |
| lbs | 375 | 441 | 507 | 617 | |
| Shipping weight1 | kg | 190 | 225 | 260 | 315 |
| lbs | 419 | 496 | 573 | 694 | |
| Max load capacity of one-piece work surface | 50kg (110 lbs) | ||||
| Max load capacity of single segmented work surfaces (per 30cm segment) | 25kg (55 lbs) | ||||
1 Export packaging dimensions and weight upon request
| Size/Width | 0.9m | 1.2m | 1.5m | 1.8m |
| Voltage (V) | 230 | |||
Frequency (Hz) |
50/60 |
|||
| Filter Specification | H14 HEPA EN 1822, 99.995% MPPS | |||
| Certification | TUV | TUV, LNE | TUV | TUV |
| Size/Width | 0.9m | 1.2m | 1.5m | 1.8m | |
| Lighting power | lux | 1000 | 929 | 1500 | 1418 |
| fc | 93 | 86 | 139 | 132 | |
| Sound pressure level (dbA) per EN 12469 | 57 | 57 | |||
| Receptacles | 2 rear wall | 4 rear wall | |||
| Service Valves | 2 on each side |
||||
| SmartPorts | Two 76mm (3") plugged cable ports, one on each wall | ||||
| Size/Width | 0.9m | 1.2m | 1.5m | 1.8m | |
| Energy consumption, operating set point2 (W) | 133 | 153 | 217 | 251 | |
| Energy consumption, Night Set-Back mode (W) | 55 | 80 | |||
| Heat output, operating mode (non-vented) (BTU/hr) | 454 | 522 | 740 | 856 | |
| Heat output, night set-back mode (lights off, non vented) (BTU/hr) | 188 | 273 | |||
| Exhaust / Inflow air volume, recirculated | (m3/h) | 296 | 395 | 493 | 592 |
| (cfm) | 174 | 232 | 290 | 348 | |
| Exhaust volume, thimble ducted | (m3/hr) | 385 | 513 | 641 | 770 |
| (cfm) | 227 | 302 | 377 | 453 | |
| Request a quote | 51032328 › | 51032329 › | 51032330 › | 51032331 › | |
2 Clean filters, fans operating at nominal set point, interior lighting activated
Key features to help you achieve your next discovery
Based on over 40 years of engineering and design breakthroughs, our expertise is represented in key features that span our portfolio. Together these enhancements provide valuable contamination control.
Thermo Scientific BSCs use separate downflow and exhaust fans. SmartFlow compensation maintains inflow and the overall balance of inflow and downflow through real-time adjustment of the exhaust fan for constant flow and comparable adjustment to downflow. Thermo Scientific 2030i has SmartFlow Plus with fully implemented independent compensation to maintain constant inflow and constant downflow.
Thermo Scientific provides a number of different options for UV disinfection. Our traditional approach with UV positioned above the window provides an even disinfection inside the work area. Our patented Crossbeam UV feature that ensures optimal coverage and reduces shadows on the work surface. And our new UV compensation feature, when selected, increases the selected UV disinfection cycle time to compensate for bulb life and assure the desired germicidal energy is delivered automatically. All of our UV options are available with programmable timers.
All Thermo Scientific BSCs provide visual and audible alarms to warn of improper window position and unacceptable variation in airflow. Thermo Scientific 2030i has a unique interface that graphically depicts the alarm condition, text that advises of the specific concern and selectable audible tone to allow easy differentiation between window position and airflow problems.
Thermo Scientific offers BSC users a choice of window/sash types between a counterbalanced "manual" sash type window or our electric sash type window for better ergonomics. In addition, the electric sash on the Herasafe 2030i has an available auto-start/stop. In Stand-by or Off, the AutoStart key is displayed and will raise the window to operating position and energize the fans, LED lights and/or internal receptacles as configured with one button. In Operation, the AutoStop key is displayed and will close the window, de-energize the fans/ LED lights/ internal receptacles and initiate UV disinfection cycle as configured by pressing one button.
Thermo Scientific BSCs allow a choice of internal surfaces including more economical partially coated interiors, type 304 stainless steel or type 316 stainless steel for greater chemical resistance. Our BSCs offer either double sided walls with negatively pressurized interstitial space for enhanced structure containment or transparent side panels for improved ergonomics. Work surfaces within the cabinet can be flat for easy cleaning or indented for containment of larger spills onto the work surface.
Thermo Scientific BSCs use separate downflow and exhaust fans with DC electric motors. This combination of energy efficient DC motors with the fan based balance greatly reduces energy consumption compared to to traditional AC motor units with mechanical balancing dampers.
Thermo Scientific BSCs allow a choice of traditional fluorescent lights for cabinet illumination or more energy efficient and sustainable LED lighting.