Search
We have collaborated with Structura Biotechnology Inc. to develop an integrated version of CryoSPARC Live™ to significantly enhance efficiency of Thermo Scientific cryo-transmission electron microscopes (cryo-TEMs). Embedded CryoSPARC Live significantly shortens time to results by providing real-time, multi-dimensional feedback on sample quality and transmission electron microscope settings. It allows you to adjust imaging and processing parameters at any time, increasing overall data quality and minimizing time wasted on poor quality samples. With the ability to fully integrate with Thermo Scientific Smart EPU Software, it can support a growing set of automations.
Embedded CryoSPARC Live is installed on microscope hardware. Our digital service team provides support through installation and maintenance, and our application specialists provide integral workflow training.
*CryoSPARC Live is a trademark of Structura Biotechnology Inc. and is used under license.
The simple guided workflow, intuitive user interface, and integration with Smart EPU Software enable usability for researchers spanning a broad range of scientific expertise.
Seamless integration with Smart EPU Software enables real-time decision making and helps you easily enact optional data acquisition strategies. Easily export Embedded CryoSPARC Live sessions for more advanced downstream processing in CryoSPARC™.
Embedded CryoSPARC Live can significantly shorten time to results:
Embedded CryoSPARC Live works flawlessly with Smart Plugins from Smart EPU Software to adjust auto processing settings, and it fully integrates with Thermo Scientific CryoFlow Software’s data management platform for automatic data exchange and improved ease of use.
For Research Use Only. Not for use in diagnostic procedures.