Ramsey Weigh Belt Feeder
Reduce waste, maintain consistency, increase profits
Precise feeding of process materials is often critical to maintaining product quality. An accurate and reliable weigh feeder can reduce material waste, help maintain blend consistency, increase profits, and meet customer specifications.
Thermo Scientific™ Ramsey™ weighbelt feeders accurately control process material feed rates. Whether it is raw material for raw mix proportioning or coal for the kiln, we have the right feeder for your application. Our loss-in-weight feeders are an excellent choice for dry, dusty, or variable-density materials.
Contact a sales representative
Quick links
Featured weighbelt feeders
Measure industrial materials at moderate feed rates with the Ramsey 90.125 Mid-Range Weighbelt Feeder. A unique cantilevered and formed channel frame design provides a sturdy weigh structure and allows for easy maintenance.
Increase sensitivity for more accurate weighing of the lightest materials with the Ramsey 90.150 Low Capacity Weighbelt Feeder. This weighbelt feeder provides a consistent flow of material and permits easy cleaning and maintenance.
Improve blending and ratio control with the Ramsey Micro-Tech 9105 Weighbelt Feeder Controller. This specialized, multi-functional digital control device incorporates all weighing, logic, and control functions into a single dedicated package.
Resources
Support
Invest in peace of mind with a Thermo Scientific service agreement. Ensure minimum downtime with known costs and priority response times. Let us tailor a plan to meet your needs. For parts, technical support, or field assistance, please call 800-445-3503.